Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33416489-33416762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUTA
Entrez Gene ID
51596
Gene Name
cutA divalent cation tolerance homolog (E. coli)
Gene Aliases
ACHAP, C6orf82
UniGene
Hs.520070
Gene Chromosome Location
Chr.6: 33416542-33418288 on Build GRCh38
Transcripts
NM_001014433.2 NM_001014837.1 NM_001014838.1 NM_001014840.1 NM_015921.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4878031 c.538C>T p.L180L Chr.6: 33416709-33416709 on Build GRCh38
4857727 c.521A>G p.Q174R Chr.6: 33416726-33416726 on Build GRCh38
3625732 c.377T>C p.I126T Chr.6: 33416744-33416744 on Build GRCh38
3625729 c.596G>A p.*199* Chr.6: 33416651-33416651 on Build GRCh38
5737566 c.584C>T p.T195I Chr.6: 33416663-33416663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777906203 Chr.6: 33416612-33416613 on Build GRCh38
rs201868307 Chr.6: 33416739-33416739 on Build GRCh38
rs763423148 Chr.6: 33416651-33416651 on Build GRCh38
rs375282900 Chr.6: 33416684-33416684 on Build GRCh38
rs540282025 Chr.6: 33416603-33416603 on Build GRCh38
rs760288288 Chr.6: 33416697-33416697 on Build GRCh38
rs748340965 Chr.6: 33416656-33416656 on Build GRCh38
rs374614248 Chr.6: 33416646-33416646 on Build GRCh38
rs867927341 Chr.6: 33416563-33416563 on Build GRCh38
rs762423401 Chr.6: 33416615-33416615 on Build GRCh38
rs1027165249 Chr.6: 33416537-33416537 on Build GRCh38
rs973848055 Chr.6: 33416722-33416722 on Build GRCh38
rs774504365 Chr.6: 33416686-33416687 on Build GRCh38
rs183773510 Chr.6: 33416532-33416532 on Build GRCh38
rs370122087 Chr.6: 33416616-33416616 on Build GRCh38
rs769140145 Chr.6: 33416612-33416612 on Build GRCh38
rs764138453 Chr.6: 33416727-33416727 on Build GRCh38
rs776048307 Chr.6: 33416708-33416708 on Build GRCh38
rs878977559 Chr.6: 33416581-33416581 on Build GRCh38
rs753472290 Chr.6: 33416635-33416635 on Build GRCh38
rs781342305 Chr.6: 33416578-33416578 on Build GRCh38
rs772678086 Chr.6: 33416691-33416691 on Build GRCh38
rs563470498 Chr.6: 33416661-33416661 on Build GRCh38
rs188454255 Chr.6: 33416556-33416556 on Build GRCh38
rs1053683 Chr.6: 33416675-33416675 on Build GRCh38
rs199741480 Chr.6: 33416687-33416687 on Build GRCh38
rs952413735 Chr.6: 33416541-33416541 on Build GRCh38
rs760074942 Chr.6: 33416617-33416617 on Build GRCh38
rs773698135 Chr.6: 33416580-33416580 on Build GRCh38
rs897320262 Chr.6: 33416517-33416517 on Build GRCh38
rs774309559 Chr.6: 33416604-33416606 on Build GRCh38
rs758264817 Chr.6: 33416575-33416575 on Build GRCh38
rs888638429 Chr.6: 33416704-33416704 on Build GRCh38
rs902607531 Chr.6: 33416525-33416525 on Build GRCh38
rs1023285359 Chr.6: 33416545-33416548 on Build GRCh38
rs766106995 Chr.6: 33416632-33416632 on Build GRCh38
rs138574267 Chr.6: 33416681-33416681 on Build GRCh38
rs752519101 Chr.6: 33416714-33416714 on Build GRCh38
rs762622672 Chr.6: 33416725-33416725 on Build GRCh38
rs745886202 Chr.6: 33416590-33416590 on Build GRCh38
rs772316615 Chr.6: 33416683-33416684 on Build GRCh38
rs200454650 Chr.6: 33416643-33416643 on Build GRCh38
rs994298867 Chr.6: 33416527-33416527 on Build GRCh38
rs371701025 Chr.6: 33416713-33416713 on Build GRCh38
rs371083164 Chr.6: 33416667-33416667 on Build GRCh38
rs377559003 Chr.6: 33416647-33416647 on Build GRCh38
rs144359180 Chr.6: 33416737-33416737 on Build GRCh38
rs768512627 Chr.6: 33416589-33416590 on Build GRCh38
rs770084680 Chr.6: 33416597-33416597 on Build GRCh38
rs1041056174 Chr.6: 33416657-33416657 on Build GRCh38
rs770339717 Chr.6: 33416700-33416700 on Build GRCh38
rs540797551 Chr.6: 33416547-33416547 on Build GRCh38
rs558572694 Chr.6: 33416544-33416545 on Build GRCh38
rs915089490 Chr.6: 33416607-33416614 on Build GRCh38
rs201145897 Chr.6: 33416666-33416666 on Build GRCh38
rs939650408 Chr.6: 33416716-33416716 on Build GRCh38
rs760806385 Chr.6: 33416725-33416726 on Build GRCh38
rs149930961 Chr.6: 33416613-33416613 on Build GRCh38
rs41267649 Chr.6: 33416696-33416696 on Build GRCh38
rs780106729 Chr.6: 33416602-33416602 on Build GRCh38
rs756136957 Chr.6: 33416652-33416652 on Build GRCh38

View Full Product Details