Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99330228-99330485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM1
Entrez Gene ID
26507
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 1
Gene Aliases
ACDP1, CLP-1
UniGene
Hs.274579
Gene Chromosome Location
Chr.10: 99329099-99394330 on Build GRCh38
Transcripts
NM_020348.2 XM_005269692.3 XM_011539629.1 XM_011539630.1 XM_011539631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631289 c.1043C>T p.S348L Chr.10: 99330430-99330430 on Build GRCh38
4165363 c.1008C>T p.F336F Chr.10: 99330395-99330395 on Build GRCh38
3433428 c.988G>A p.V330M Chr.10: 99330375-99330375 on Build GRCh38
6551232 c.1045C>T p.R349W Chr.10: 99330432-99330432 on Build GRCh38
4846744 c.1062C>T p.I354I Chr.10: 99330449-99330449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375175318 Chr.10: 99330460-99330460 on Build GRCh38
rs759656876 Chr.10: 99330303-99330303 on Build GRCh38
rs767922278 Chr.10: 99330428-99330428 on Build GRCh38
rs554656536 Chr.10: 99330288-99330288 on Build GRCh38
rs781664354 Chr.10: 99330319-99330319 on Build GRCh38
rs869025226 Chr.10: 99330344-99330344 on Build GRCh38
rs751849944 Chr.10: 99330414-99330414 on Build GRCh38
rs933446502 Chr.10: 99330383-99330383 on Build GRCh38
rs776688198 Chr.10: 99330376-99330376 on Build GRCh38
rs746418618 Chr.10: 99330324-99330324 on Build GRCh38
rs921598176 Chr.10: 99330437-99330437 on Build GRCh38
rs758580754 Chr.10: 99330306-99330306 on Build GRCh38
rs372555911 Chr.10: 99330305-99330305 on Build GRCh38
rs776776118 Chr.10: 99330302-99330302 on Build GRCh38
rs773055975 Chr.10: 99330359-99330359 on Build GRCh38
rs867369746 Chr.10: 99330454-99330454 on Build GRCh38
rs781404422 Chr.10: 99330410-99330413 on Build GRCh38
rs764212142 Chr.10: 99330406-99330406 on Build GRCh38
rs747766715 Chr.10: 99330351-99330351 on Build GRCh38
rs764454976 Chr.10: 99330308-99330308 on Build GRCh38
rs895244565 Chr.10: 99330354-99330354 on Build GRCh38
rs757680050 Chr.10: 99330315-99330315 on Build GRCh38
rs905646110 Chr.10: 99330264-99330264 on Build GRCh38
rs763179973 Chr.10: 99330404-99330404 on Build GRCh38
rs572699738 Chr.10: 99330338-99330338 on Build GRCh38
rs771856848 Chr.10: 99330269-99330269 on Build GRCh38
rs377197002 Chr.10: 99330311-99330311 on Build GRCh38
rs756580417 Chr.10: 99330466-99330466 on Build GRCh38
rs760630170 Chr.10: 99330271-99330271 on Build GRCh38
rs757259018 Chr.10: 99330276-99330279 on Build GRCh38
rs750886240 Chr.10: 99330431-99330431 on Build GRCh38
rs988035344 Chr.10: 99330384-99330384 on Build GRCh38
rs770888746 Chr.10: 99330371-99330371 on Build GRCh38
rs772885251 Chr.10: 99330270-99330270 on Build GRCh38
rs771633671 Chr.10: 99330353-99330353 on Build GRCh38
rs765283165 Chr.10: 99330304-99330304 on Build GRCh38
rs766405339 Chr.10: 99330281-99330281 on Build GRCh38
rs1012313922 Chr.10: 99330375-99330375 on Build GRCh38
rs765320855 Chr.10: 99330382-99330382 on Build GRCh38
rs370414041 Chr.10: 99330337-99330337 on Build GRCh38
rs759713178 Chr.10: 99330279-99330279 on Build GRCh38
rs78082541 Chr.10: 99330294-99330294 on Build GRCh38
rs540244284 Chr.10: 99330392-99330392 on Build GRCh38
rs200388932 Chr.10: 99330366-99330366 on Build GRCh38
rs1003107195 Chr.10: 99330272-99330272 on Build GRCh38
rs759568931 Chr.10: 99330379-99330379 on Build GRCh38
rs952155877 Chr.10: 99330388-99330388 on Build GRCh38
rs868789701 Chr.10: 99330449-99330449 on Build GRCh38
rs950689712 Chr.10: 99330283-99330283 on Build GRCh38
rs757590337 Chr.10: 99330422-99330422 on Build GRCh38

View Full Product Details