Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29999705-29999966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB5
Entrez Gene ID
374786
Gene Name
EF-hand calcium binding domain 5
Gene Aliases
-
UniGene
Hs.662411
Gene Chromosome Location
Chr.17: 29929856-30108452 on Build GRCh38
Transcripts
NM_001145053.1 NM_198529.3 NR_026738.1 XM_011524759.1 XM_011524763.2 XM_011524765.1 XM_011524766.2 XM_011524768.2 XM_017024591.1 XM_017024592.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
977243 c.1016_1017insA p.R340fs*16 Chr.17: 29999948-29999949 on Build GRCh38
5448962 c.823C>T p.L275L Chr.17: 29999923-29999923 on Build GRCh38
5448963 c.991C>T p.L331L Chr.17: 29999923-29999923 on Build GRCh38
6409667 c.977C>A p.S326* Chr.17: 29999909-29999909 on Build GRCh38
6409666 c.809C>A p.S270* Chr.17: 29999909-29999909 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs543464918 Chr.17: 29999767-29999767 on Build GRCh38
rs766332751 Chr.17: 29999861-29999861 on Build GRCh38
rs972388186 Chr.17: 29999919-29999919 on Build GRCh38
rs922271916 Chr.17: 29999748-29999748 on Build GRCh38
rs189325863 Chr.17: 29999916-29999916 on Build GRCh38
rs540803734 Chr.17: 29999903-29999903 on Build GRCh38
rs767133273 Chr.17: 29999729-29999729 on Build GRCh38
rs1021335445 Chr.17: 29999739-29999739 on Build GRCh38
rs961237397 Chr.17: 29999906-29999906 on Build GRCh38
rs959195786 Chr.17: 29999855-29999855 on Build GRCh38
rs267604789 Chr.17: 29999930-29999930 on Build GRCh38
rs752724038 Chr.17: 29999839-29999839 on Build GRCh38
rs1033541244 Chr.17: 29999840-29999840 on Build GRCh38
rs760126508 Chr.17: 29999857-29999857 on Build GRCh38
rs184227825 Chr.17: 29999870-29999870 on Build GRCh38
rs1010166141 Chr.17: 29999864-29999864 on Build GRCh38
rs115160874 Chr.17: 29999791-29999791 on Build GRCh38
rs1024204356 Chr.17: 29999917-29999917 on Build GRCh38
rs984036652 Chr.17: 29999881-29999881 on Build GRCh38
rs760669059 Chr.17: 29999852-29999852 on Build GRCh38
rs558523887 Chr.17: 29999868-29999868 on Build GRCh38
rs893848051 Chr.17: 29999727-29999727 on Build GRCh38
rs889521646 Chr.17: 29999812-29999812 on Build GRCh38
rs759491606 Chr.17: 29999927-29999927 on Build GRCh38
rs935873209 Chr.17: 29999827-29999827 on Build GRCh38
rs774764288 Chr.17: 29999869-29999872 on Build GRCh38
rs866552038 Chr.17: 29999939-29999939 on Build GRCh38
rs752473747 Chr.17: 29999936-29999936 on Build GRCh38
rs1054041327 Chr.17: 29999724-29999724 on Build GRCh38
rs970856417 Chr.17: 29999766-29999766 on Build GRCh38
rs909810736 Chr.17: 29999888-29999888 on Build GRCh38

View Full Product Details