Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 47142982-47143229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
UniGene
Hs.472877
Gene Chromosome Location
Chr.20: 46894624-47188844 on Build GRCh38
Transcripts
NM_005244.4 NM_172110.3 XM_005260327.2 XM_017027721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6840062 c.978+1G>A p.? Chr.20: 47143149-47143149 on Build GRCh38
1681736 c.893C>T p.T298I Chr.20: 47143063-47143063 on Build GRCh38
6558849 c.904G>T p.V302L Chr.20: 47143074-47143074 on Build GRCh38
1740684 c.933G>T p.M311I Chr.20: 47143103-47143103 on Build GRCh38
6558848 c.903C>T p.S301S Chr.20: 47143073-47143073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021852897 Chr.20: 47143193-47143193 on Build GRCh38
rs745988411 Chr.20: 47143020-47143020 on Build GRCh38
rs771650179 Chr.20: 47143179-47143179 on Build GRCh38
rs944132387 Chr.20: 47143097-47143097 on Build GRCh38
rs773129423 Chr.20: 47143043-47143043 on Build GRCh38
rs780109158 Chr.20: 47143078-47143078 on Build GRCh38
rs747299291 Chr.20: 47143080-47143080 on Build GRCh38
rs778414856 Chr.20: 47143009-47143009 on Build GRCh38
rs762936646 Chr.20: 47143049-47143049 on Build GRCh38
rs749085280 Chr.20: 47143100-47143100 on Build GRCh38
rs200269249 Chr.20: 47143070-47143070 on Build GRCh38
rs3818915 Chr.20: 47143023-47143023 on Build GRCh38
rs770732276 Chr.20: 47143138-47143138 on Build GRCh38
rs758943992 Chr.20: 47143173-47143173 on Build GRCh38
rs111303908 Chr.20: 47143069-47143069 on Build GRCh38
rs915577727 Chr.20: 47143024-47143024 on Build GRCh38
rs757385820 Chr.20: 47143013-47143013 on Build GRCh38
rs539723989 Chr.20: 47143004-47143004 on Build GRCh38
rs779509858 Chr.20: 47143073-47143073 on Build GRCh38
rs141317409 Chr.20: 47143066-47143066 on Build GRCh38
rs146794889 Chr.20: 47143133-47143133 on Build GRCh38
rs774366276 Chr.20: 47143153-47143153 on Build GRCh38
rs558133381 Chr.20: 47143008-47143008 on Build GRCh38
rs777486517 Chr.20: 47143094-47143094 on Build GRCh38
rs762264147 Chr.20: 47143198-47143198 on Build GRCh38
rs899850096 Chr.20: 47143185-47143185 on Build GRCh38
rs770914435 Chr.20: 47143052-47143052 on Build GRCh38
rs747918390 Chr.20: 47143026-47143026 on Build GRCh38
rs368297287 Chr.20: 47143077-47143077 on Build GRCh38
rs780335186 Chr.20: 47143025-47143025 on Build GRCh38
rs775273072 Chr.20: 47143183-47143183 on Build GRCh38
rs1041475306 Chr.20: 47143147-47143147 on Build GRCh38
rs752443976 Chr.20: 47143055-47143055 on Build GRCh38
rs769564846 Chr.20: 47143089-47143089 on Build GRCh38
rs374388625 Chr.20: 47143195-47143195 on Build GRCh38
rs141609454 Chr.20: 47143191-47143191 on Build GRCh38
rs749880445 Chr.20: 47143010-47143010 on Build GRCh38
rs757724776 Chr.20: 47143171-47143171 on Build GRCh38
rs537062071 Chr.20: 47143054-47143054 on Build GRCh38
rs767857152 Chr.20: 47143031-47143032 on Build GRCh38
rs370808552 Chr.20: 47143017-47143017 on Build GRCh38
rs769618216 Chr.20: 47143028-47143028 on Build GRCh38

View Full Product Details