Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26444129-26444402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRC1
Entrez Gene ID
92749
Gene Name
dynein regulatory complex subunit 1
Gene Aliases
C2orf39, CCDC164, CILD21
UniGene
Hs.393714
Gene Chromosome Location
Chr.2: 26401912-26456713 on Build GRCh38
Transcripts
NM_145038.4 XM_005264637.3 XM_005264638.3 XM_017005271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6091076 c.1080G>T p.K360N Chr.2: 26444273-26444273 on Build GRCh38
7285279 c.1111T>C p.S371P Chr.2: 26444304-26444304 on Build GRCh38
5973731 c.1050T>C p.N350N Chr.2: 26444243-26444243 on Build GRCh38
5396734 c.1036G>A p.D346N Chr.2: 26444229-26444229 on Build GRCh38
1019358 c.1158C>A p.A386A Chr.2: 26444351-26444351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765070685 Chr.2: 26444281-26444283 on Build GRCh38
rs115138591 Chr.2: 26444154-26444154 on Build GRCh38
rs377027994 Chr.2: 26444178-26444178 on Build GRCh38
rs947681352 Chr.2: 26444334-26444334 on Build GRCh38
rs755740597 Chr.2: 26444294-26444294 on Build GRCh38
rs773498075 Chr.2: 26444330-26444330 on Build GRCh38
rs1050722572 Chr.2: 26444180-26444180 on Build GRCh38
rs747821772 Chr.2: 26444175-26444175 on Build GRCh38
rs755494118 Chr.2: 26444263-26444263 on Build GRCh38
rs762125120 Chr.2: 26444236-26444236 on Build GRCh38
rs777356236 Chr.2: 26444302-26444302 on Build GRCh38
rs1038144571 Chr.2: 26444172-26444172 on Build GRCh38
rs763083128 Chr.2: 26444259-26444259 on Build GRCh38
rs778301620 Chr.2: 26444307-26444307 on Build GRCh38
rs533255445 Chr.2: 26444368-26444368 on Build GRCh38
rs765601461 Chr.2: 26444244-26444244 on Build GRCh38
rs529070805 Chr.2: 26444310-26444310 on Build GRCh38
rs768598799 Chr.2: 26444322-26444322 on Build GRCh38
rs145445723 Chr.2: 26444306-26444306 on Build GRCh38
rs745800344 Chr.2: 26444221-26444221 on Build GRCh38
rs763278269 Chr.2: 26444359-26444359 on Build GRCh38
rs916455994 Chr.2: 26444272-26444272 on Build GRCh38
rs145305515 Chr.2: 26444316-26444316 on Build GRCh38
rs1053432675 Chr.2: 26444246-26444246 on Build GRCh38
rs928722428 Chr.2: 26444297-26444297 on Build GRCh38
rs375562110 Chr.2: 26444185-26444185 on Build GRCh38
rs78892546 Chr.2: 26444260-26444260 on Build GRCh38
rs3795959 Chr.2: 26444376-26444376 on Build GRCh38
rs771972894 Chr.2: 26444311-26444311 on Build GRCh38
rs368859380 Chr.2: 26444305-26444305 on Build GRCh38
rs773586230 Chr.2: 26444258-26444258 on Build GRCh38
rs771844801 Chr.2: 26444225-26444225 on Build GRCh38
rs141347582 Chr.2: 26444317-26444317 on Build GRCh38
rs761874729 Chr.2: 26444172-26444173 on Build GRCh38
rs184506507 Chr.2: 26444283-26444283 on Build GRCh38
rs970137380 Chr.2: 26444275-26444275 on Build GRCh38
rs3795958 Chr.2: 26444262-26444262 on Build GRCh38
rs775210361 Chr.2: 26444227-26444227 on Build GRCh38
rs909455212 Chr.2: 26444371-26444371 on Build GRCh38
rs35094910 Chr.2: 26444286-26444286 on Build GRCh38
rs774306956 Chr.2: 26444188-26444188 on Build GRCh38
rs200534192 Chr.2: 26444177-26444177 on Build GRCh38

View Full Product Details