Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75050804-75051047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC2
Entrez Gene ID
3747
Gene Name
potassium voltage-gated channel subfamily C member 2
Gene Aliases
KV3.2
UniGene
Hs.27214
Gene Chromosome Location
Chr.12: 75040078-75209815 on Build GRCh38
Transcripts
NM_001260497.1 NM_001260498.1 NM_001260499.1 NM_139136.3 NM_139137.3 NM_153748.2 XM_005268855.3 XM_006719382.3 XM_006719383.3 XM_006719384.3 XM_006719385.3 XM_017019282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603833 c.1114G>A p.G372R Chr.12: 75050891-75050891 on Build GRCh38
5667983 c.1074T>C p.I358I Chr.12: 75050931-75050931 on Build GRCh38
7191307 c.980A>G p.Y327C Chr.12: 75051025-75051025 on Build GRCh38
6073580 c.958T>C p.F320L Chr.12: 75051047-75051047 on Build GRCh38
4460396 c.1164C>T p.F388F Chr.12: 75050841-75050841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149433460 Chr.12: 75050859-75050859 on Build GRCh38
rs763247148 Chr.12: 75050923-75050923 on Build GRCh38
rs199659211 Chr.12: 75050997-75050997 on Build GRCh38
rs377231149 Chr.12: 75050976-75050976 on Build GRCh38
rs550561919 Chr.12: 75050917-75050917 on Build GRCh38
rs200433539 Chr.12: 75050984-75050984 on Build GRCh38
rs201237451 Chr.12: 75051015-75051015 on Build GRCh38
rs560846232 Chr.12: 75050919-75050919 on Build GRCh38
rs529762229 Chr.12: 75050990-75050990 on Build GRCh38
rs372364104 Chr.12: 75051003-75051003 on Build GRCh38
rs1031184645 Chr.12: 75050939-75050939 on Build GRCh38
rs772977335 Chr.12: 75050921-75050921 on Build GRCh38
rs199857245 Chr.12: 75050895-75050895 on Build GRCh38
rs148618018 Chr.12: 75050909-75050909 on Build GRCh38
rs750183803 Chr.12: 75050864-75050864 on Build GRCh38
rs972000837 Chr.12: 75050841-75050841 on Build GRCh38
rs774800739 Chr.12: 75050933-75050933 on Build GRCh38
rs768237525 Chr.12: 75050906-75050906 on Build GRCh38
rs760061175 Chr.12: 75050970-75050970 on Build GRCh38
rs139012036 Chr.12: 75050838-75050838 on Build GRCh38
rs771289771 Chr.12: 75050925-75050925 on Build GRCh38
rs754124018 Chr.12: 75051024-75051024 on Build GRCh38
rs200951837 Chr.12: 75050879-75050879 on Build GRCh38
rs143847287 Chr.12: 75050886-75050886 on Build GRCh38
rs374596117 Chr.12: 75050888-75050888 on Build GRCh38

View Full Product Details