Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107944978-107945249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6448934 c.601G>C p.E201Q Chr.4: 107945080-107945080 on Build GRCh38
2953276 c.603G>T p.E201D Chr.4: 107945082-107945082 on Build GRCh38
3824922 c.665C>T p.S222F Chr.4: 107945144-107945144 on Build GRCh38
7331046 c.693C>T p.I231I Chr.4: 107945172-107945172 on Build GRCh38
2953279 c.680C>T p.A227V Chr.4: 107945159-107945159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021754830 Chr.4: 107945031-107945031 on Build GRCh38
rs755611981 Chr.4: 107945205-107945205 on Build GRCh38
rs746319505 Chr.4: 107945036-107945036 on Build GRCh38
rs775537061 Chr.4: 107945159-107945159 on Build GRCh38
rs757598063 Chr.4: 107945030-107945030 on Build GRCh38
rs752782576 Chr.4: 107944999-107944999 on Build GRCh38
rs765631121 Chr.4: 107945175-107945175 on Build GRCh38
rs200439778 Chr.4: 107945008-107945008 on Build GRCh38
rs942548642 Chr.4: 107945220-107945220 on Build GRCh38
rs1010581109 Chr.4: 107945021-107945021 on Build GRCh38
rs149763668 Chr.4: 107945161-107945161 on Build GRCh38
rs748925272 Chr.4: 107945115-107945115 on Build GRCh38
rs758958727 Chr.4: 107945191-107945191 on Build GRCh38
rs757429377 Chr.4: 107945164-107945164 on Build GRCh38
rs779164469 Chr.4: 107945035-107945035 on Build GRCh38
rs1057519146 Chr.4: 107945033-107945033 on Build GRCh38
rs747717076 Chr.4: 107945050-107945050 on Build GRCh38
rs539068584 Chr.4: 107945119-107945119 on Build GRCh38
rs748913829 Chr.4: 107945216-107945216 on Build GRCh38
rs754117744 Chr.4: 107945023-107945023 on Build GRCh38
rs778117975 Chr.4: 107945019-107945019 on Build GRCh38
rs769311510 Chr.4: 107945107-107945107 on Build GRCh38
rs756288347 Chr.4: 107945006-107945006 on Build GRCh38
rs761764079 Chr.4: 107945197-107945197 on Build GRCh38
rs772710211 Chr.4: 107945046-107945046 on Build GRCh38
rs139692261 Chr.4: 107945150-107945150 on Build GRCh38
rs759406615 Chr.4: 107945146-107945146 on Build GRCh38
rs750883867 Chr.4: 107945184-107945184 on Build GRCh38
rs143913941 Chr.4: 107945108-107945108 on Build GRCh38
rs147256952 Chr.4: 107945118-107945118 on Build GRCh38
rs972710866 Chr.4: 107945144-107945144 on Build GRCh38
rs145638445 Chr.4: 107945198-107945198 on Build GRCh38
rs774149338 Chr.4: 107945127-107945127 on Build GRCh38
rs754039267 Chr.4: 107945163-107945163 on Build GRCh38
rs760847189 Chr.4: 107945160-107945160 on Build GRCh38
rs777170829 Chr.4: 107945211-107945211 on Build GRCh38
rs960344604 Chr.4: 107945123-107945123 on Build GRCh38
rs780843477 Chr.4: 107945049-107945049 on Build GRCh38

View Full Product Details