Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 146029698-146029927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM1
Entrez Gene ID
2911
Gene Name
glutamate metabotropic receptor 1
Gene Aliases
GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13
UniGene
Hs.32945
Gene Chromosome Location
Chr.6: 146027634-146437598 on Build GRCh38
Transcripts
NM_001278064.1 NM_001278065.1 NM_001278066.1 NM_001278067.1 XM_011535782.1 XM_017010783.1 XM_017010784.1 XM_017010785.1 XM_017010786.1 XM_017010787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3858862 c.291C>A p.P97P Chr.6: 146029808-146029808 on Build GRCh38
3017760 c.339C>T p.S113S Chr.6: 146029856-146029856 on Build GRCh38
3017748 c.190G>A p.E64K Chr.6: 146029707-146029707 on Build GRCh38
7281766 c.194G>T p.R65M Chr.6: 146029711-146029711 on Build GRCh38
6529984 c.330G>C p.W110C Chr.6: 146029847-146029847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772338184 Chr.6: 146029814-146029814 on Build GRCh38
rs922244236 Chr.6: 146029766-146029766 on Build GRCh38
rs762000484 Chr.6: 146029789-146029789 on Build GRCh38
rs749347616 Chr.6: 146029760-146029760 on Build GRCh38
rs779134688 Chr.6: 146029893-146029893 on Build GRCh38
rs781271598 Chr.6: 146029856-146029856 on Build GRCh38
rs748392238 Chr.6: 146029768-146029768 on Build GRCh38
rs554574441 Chr.6: 146029734-146029734 on Build GRCh38
rs140675167 Chr.6: 146029754-146029754 on Build GRCh38
rs777166317 Chr.6: 146029795-146029795 on Build GRCh38
rs34559510 Chr.6: 146029839-146029840 on Build GRCh38
rs145861054 Chr.6: 146029781-146029781 on Build GRCh38
rs772412659 Chr.6: 146029777-146029777 on Build GRCh38
rs771323107 Chr.6: 146029790-146029790 on Build GRCh38
rs755976570 Chr.6: 146029837-146029837 on Build GRCh38
rs543481007 Chr.6: 146029797-146029797 on Build GRCh38
rs779226659 Chr.6: 146029767-146029767 on Build GRCh38
rs202052621 Chr.6: 146029838-146029838 on Build GRCh38
rs939211361 Chr.6: 146029810-146029810 on Build GRCh38
rs748302318 Chr.6: 146029898-146029898 on Build GRCh38
rs374804458 Chr.6: 146029723-146029723 on Build GRCh38
rs750173407 Chr.6: 146029826-146029826 on Build GRCh38
rs753847166 Chr.6: 146029879-146029879 on Build GRCh38
rs758531938 Chr.6: 146029901-146029901 on Build GRCh38
rs751246579 Chr.6: 146029722-146029722 on Build GRCh38
rs762412199 Chr.6: 146029796-146029796 on Build GRCh38
rs767268356 Chr.6: 146029823-146029823 on Build GRCh38
rs969452007 Chr.6: 146029758-146029758 on Build GRCh38
rs755049118 Chr.6: 146029890-146029890 on Build GRCh38
rs112221974 Chr.6: 146029727-146029727 on Build GRCh38
rs780098207 Chr.6: 146029757-146029757 on Build GRCh38
rs746101221 Chr.6: 146029878-146029879 on Build GRCh38
rs768886884 Chr.6: 146029762-146029762 on Build GRCh38
rs557036170 Chr.6: 146029809-146029809 on Build GRCh38

View Full Product Details