Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 210099891-210100130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT14
Entrez Gene ID
255928
Gene Name
synaptotagmin 14
Gene Aliases
SCAR11, sytXIV
UniGene
Hs.658866
Gene Chromosome Location
Chr.1: 209938165-210171315 on Build GRCh38
Transcripts
NM_001146261.2 NM_001146262.2 NM_001146264.2 NM_001256006.1 NM_153262.3 NR_027459.2 XM_006711262.3 XM_017000931.1 XM_017000932.1 XM_017000933.1 XM_017000934.1 XM_017000935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180098 c.774A>G p.E258E Chr.1: 210100071-210100071 on Build GRCh38
3482957 c.719T>G p.M240R Chr.1: 210100016-210100016 on Build GRCh38
233531 c.796G>A p.V266M Chr.1: 210100093-210100093 on Build GRCh38
6534200 c.809A>G p.Y270C Chr.1: 210100106-210100106 on Build GRCh38
4230526 c.812A>G p.D271G Chr.1: 210100109-210100109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375826096 Chr.1: 210100037-210100037 on Build GRCh38
rs1042584123 Chr.1: 210100085-210100085 on Build GRCh38
rs751860484 Chr.1: 210100013-210100013 on Build GRCh38
rs140363425 Chr.1: 210099991-210099991 on Build GRCh38
rs138599742 Chr.1: 210100040-210100040 on Build GRCh38
rs771266667 Chr.1: 210099987-210099987 on Build GRCh38
rs371207931 Chr.1: 210100004-210100004 on Build GRCh38
rs774224437 Chr.1: 210099961-210099961 on Build GRCh38
rs749255767 Chr.1: 210100071-210100071 on Build GRCh38
rs759630641 Chr.1: 210099996-210099996 on Build GRCh38
rs1054056952 Chr.1: 210099929-210099929 on Build GRCh38
rs199646737 Chr.1: 210100062-210100062 on Build GRCh38
rs534094479 Chr.1: 210100009-210100009 on Build GRCh38
rs765711998 Chr.1: 210100003-210100003 on Build GRCh38
rs760994552 Chr.1: 210099985-210099985 on Build GRCh38
rs142142387 Chr.1: 210100012-210100012 on Build GRCh38
rs755675630 Chr.1: 210100048-210100048 on Build GRCh38
rs756958681 Chr.1: 210100022-210100022 on Build GRCh38
rs370207388 Chr.1: 210100038-210100038 on Build GRCh38
rs767415863 Chr.1: 210100056-210100061 on Build GRCh38

View Full Product Details