Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109392759-109393012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL6
Entrez Gene ID
285755
Gene Name
peptidylprolyl isomerase like 6
Gene Aliases
PPIase, RSPH12, bA425D10.6, dJ919F19.1
UniGene
Hs.32234
Gene Chromosome Location
Chr.6: 109390215-109441171 on Build GRCh38
Transcripts
NM_001111298.2 NM_001286360.1 NM_001286361.1 NM_173672.4 NR_104429.1 XM_011535765.2 XM_017010774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1071961 c.926C>A p.P309H Chr.6: 109392836-109392836 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756155082 Chr.6: 109392814-109392814 on Build GRCh38
rs143629937 Chr.6: 109392859-109392859 on Build GRCh38
rs746226204 Chr.6: 109392856-109392856 on Build GRCh38
rs771244312 Chr.6: 109392817-109392817 on Build GRCh38
rs757607286 Chr.6: 109392974-109392974 on Build GRCh38
rs895503713 Chr.6: 109392945-109392945 on Build GRCh38
rs368877007 Chr.6: 109392788-109392788 on Build GRCh38
rs774918934 Chr.6: 109392842-109392842 on Build GRCh38
rs749372095 Chr.6: 109392971-109392971 on Build GRCh38
rs763608879 Chr.6: 109392891-109392891 on Build GRCh38
rs780027876 Chr.6: 109392807-109392807 on Build GRCh38
rs1045303953 Chr.6: 109392884-109392884 on Build GRCh38
rs761495611 Chr.6: 109392918-109392918 on Build GRCh38
rs760133808 Chr.6: 109392888-109392888 on Build GRCh38
rs199994620 Chr.6: 109392800-109392800 on Build GRCh38
rs775997628 Chr.6: 109392863-109392863 on Build GRCh38
rs758158097 Chr.6: 109392929-109392929 on Build GRCh38
rs764866784 Chr.6: 109392920-109392920 on Build GRCh38
rs749474726 Chr.6: 109392815-109392815 on Build GRCh38
rs766235350 Chr.6: 109392936-109392936 on Build GRCh38
rs79224538 Chr.6: 109392967-109392967 on Build GRCh38
rs995649225 Chr.6: 109392852-109392852 on Build GRCh38
rs148088638 Chr.6: 109392927-109392927 on Build GRCh38
rs372208795 Chr.6: 109392797-109392797 on Build GRCh38
rs751477946 Chr.6: 109392951-109392951 on Build GRCh38
rs756173513 Chr.6: 109392955-109392955 on Build GRCh38
rs776099194 Chr.6: 109392902-109392902 on Build GRCh38
rs887366189 Chr.6: 109392905-109392905 on Build GRCh38
rs906820403 Chr.6: 109392919-109392919 on Build GRCh38
rs771152324 Chr.6: 109392933-109392933 on Build GRCh38

View Full Product Details