Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21685169-21685438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
UniGene
Hs.408336
Gene Chromosome Location
Chr.16: 21663857-21771468 on Build GRCh38
Transcripts
NM_001161683.1 NM_144672.3 XM_011545747.2 XM_017022950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6735959 c.394G>T p.G132C Chr.16: 21685356-21685356 on Build GRCh38
4710494 c.305A>G p.Q102R Chr.16: 21685267-21685267 on Build GRCh38
6735957 c.394G>T p.G132C Chr.16: 21685356-21685356 on Build GRCh38
4710495 c.305A>G p.Q102R Chr.16: 21685267-21685267 on Build GRCh38
3794680 c.379G>C p.E127Q Chr.16: 21685341-21685341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767421718 Chr.16: 21685239-21685239 on Build GRCh38
rs370853687 Chr.16: 21685216-21685216 on Build GRCh38
rs758593584 Chr.16: 21685297-21685297 on Build GRCh38
rs776587492 Chr.16: 21685245-21685245 on Build GRCh38
rs946249025 Chr.16: 21685394-21685394 on Build GRCh38
rs963348134 Chr.16: 21685318-21685318 on Build GRCh38
rs772053837 Chr.16: 21685284-21685301 on Build GRCh38
rs751909775 Chr.16: 21685304-21685304 on Build GRCh38
rs181276948 Chr.16: 21685383-21685383 on Build GRCh38
rs772099935 Chr.16: 21685339-21685339 on Build GRCh38
rs183719341 Chr.16: 21685203-21685203 on Build GRCh38
rs755756165 Chr.16: 21685313-21685313 on Build GRCh38
rs776958501 Chr.16: 21685347-21685349 on Build GRCh38
rs367679697 Chr.16: 21685355-21685355 on Build GRCh38
rs778917245 Chr.16: 21685253-21685254 on Build GRCh38
rs777109473 Chr.16: 21685248-21685259 on Build GRCh38
rs777388246 Chr.16: 21685317-21685317 on Build GRCh38
rs373886351 Chr.16: 21685204-21685204 on Build GRCh38
rs773060183 Chr.16: 21685221-21685221 on Build GRCh38
rs376188519 Chr.16: 21685218-21685218 on Build GRCh38
rs781684262 Chr.16: 21685309-21685309 on Build GRCh38
rs750680215 Chr.16: 21685293-21685293 on Build GRCh38
rs147717988 Chr.16: 21685358-21685358 on Build GRCh38
rs202231788 Chr.16: 21685195-21685195 on Build GRCh38
rs915178636 Chr.16: 21685382-21685382 on Build GRCh38
rs774174526 Chr.16: 21685235-21685235 on Build GRCh38
rs766553207 Chr.16: 21685402-21685402 on Build GRCh38
rs748946015 Chr.16: 21685224-21685224 on Build GRCh38
rs745649295 Chr.16: 21685268-21685268 on Build GRCh38
rs766671363 Chr.16: 21685301-21685301 on Build GRCh38
rs780594618 Chr.16: 21685212-21685212 on Build GRCh38
rs770892393 Chr.16: 21685228-21685228 on Build GRCh38
rs778605976 Chr.16: 21685341-21685341 on Build GRCh38
rs748655989 Chr.16: 21685310-21685310 on Build GRCh38
rs368456979 Chr.16: 21685266-21685266 on Build GRCh38
rs747516934 Chr.16: 21685205-21685205 on Build GRCh38
rs903287616 Chr.16: 21685193-21685193 on Build GRCh38
rs765295123 Chr.16: 21685280-21685280 on Build GRCh38
rs552351972 Chr.16: 21685208-21685208 on Build GRCh38
rs770719696 Chr.16: 21685250-21685251 on Build GRCh38
rs760573914 Chr.16: 21685363-21685363 on Build GRCh38
rs755591042 Chr.16: 21685203-21685203 on Build GRCh38
rs769907669 Chr.16: 21685377-21685377 on Build GRCh38
rs748854516 Chr.16: 21685248-21685249 on Build GRCh38
rs759414633 Chr.16: 21685236-21685236 on Build GRCh38
rs755447331 Chr.16: 21685305-21685305 on Build GRCh38
rs748997472 Chr.16: 21685320-21685320 on Build GRCh38
rs981884001 Chr.16: 21685333-21685333 on Build GRCh38
rs761937183 Chr.16: 21685272-21685272 on Build GRCh38
rs913427312 Chr.16: 21685388-21685388 on Build GRCh38
rs1016182526 Chr.16: 21685295-21685295 on Build GRCh38
rs770570982 Chr.16: 21685337-21685337 on Build GRCh38

View Full Product Details