Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58503783-58504016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDRG4
Entrez Gene ID
65009
Gene Name
NDRG family member 4
Gene Aliases
BDM1, SMAP-8, SMAP8
UniGene
Hs.322430
Gene Chromosome Location
Chr.16: 58463645-58515403 on Build GRCh38
Transcripts
NM_001130487.1 NM_001242833.1 NM_001242834.1 NM_001242835.1 NM_001242836.1 NM_020465.3 NM_022910.3 NR_040072.1 XM_006721252.3 XM_006721253.3 XM_006721254.3 XM_006721256.3 XM_006721257.3 XM_006721258.3 XM_006721259.3 XM_006721260.3 XM_006721261.3 XM_006721262.3 XM_011523290.2 XM_011523291.2 XM_011523292.2 XM_011523293.2 XM_011523294.2 XM_017023582.1 XM_017023583.1 XM_017023584.1 XM_017023585.1 XM_017023586.1 XM_017023587.1 XM_017023588.1 XM_017023589.1 XM_017023590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144693 c.104C>A p.T35N Chr.16: 58503880-58503880 on Build GRCh38
7212560 c.185A>G p.D62G Chr.16: 58503805-58503805 on Build GRCh38
6144697 c.67C>A p.R23R Chr.16: 58503843-58503843 on Build GRCh38
6144691 c.260C>A p.T87N Chr.16: 58503880-58503880 on Build GRCh38
6580989 c.170C>T p.S57F Chr.16: 58503850-58503850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571308077 Chr.16: 58503985-58503985 on Build GRCh38
rs768136996 Chr.16: 58503943-58503943 on Build GRCh38
rs547018340 Chr.16: 58503970-58503971 on Build GRCh38
rs764374295 Chr.16: 58503941-58503942 on Build GRCh38
rs760170181 Chr.16: 58503966-58503966 on Build GRCh38
rs749351617 Chr.16: 58503822-58503822 on Build GRCh38
rs750911191 Chr.16: 58503819-58503819 on Build GRCh38
rs745671538 Chr.16: 58503923-58503923 on Build GRCh38
rs375101369 Chr.16: 58503903-58503903 on Build GRCh38
rs148884288 Chr.16: 58503844-58503844 on Build GRCh38
rs531493705 Chr.16: 58503904-58503904 on Build GRCh38
rs777869296 Chr.16: 58503973-58503973 on Build GRCh38
rs773896195 Chr.16: 58503944-58503944 on Build GRCh38
rs758733097 Chr.16: 58503910-58503910 on Build GRCh38
rs202237732 Chr.16: 58503864-58503864 on Build GRCh38
rs779858475 Chr.16: 58503930-58503930 on Build GRCh38
rs2427787 Chr.16: 58503993-58503993 on Build GRCh38
rs145564006 Chr.16: 58503843-58503843 on Build GRCh38
rs767840538 Chr.16: 58503876-58503876 on Build GRCh38
rs766812164 Chr.16: 58503897-58503897 on Build GRCh38
rs901536229 Chr.16: 58503848-58503848 on Build GRCh38
rs757619066 Chr.16: 58503810-58503810 on Build GRCh38
rs779283723 Chr.16: 58503826-58503826 on Build GRCh38
rs765376417 Chr.16: 58503967-58503967 on Build GRCh38
rs981942542 Chr.16: 58503946-58503946 on Build GRCh38
rs758643013 Chr.16: 58503971-58503971 on Build GRCh38
rs200710260 Chr.16: 58503865-58503865 on Build GRCh38
rs551553249 Chr.16: 58503994-58503994 on Build GRCh38
rs750787421 Chr.16: 58503890-58503890 on Build GRCh38
rs1025168153 Chr.16: 58503976-58503976 on Build GRCh38
rs771737366 Chr.16: 58503952-58503952 on Build GRCh38
rs749154966 Chr.16: 58503975-58503975 on Build GRCh38
rs527321934 Chr.16: 58503856-58503856 on Build GRCh38
rs747496947 Chr.16: 58503823-58503823 on Build GRCh38
rs769713546 Chr.16: 58503926-58503926 on Build GRCh38
rs200025649 Chr.16: 58503893-58503893 on Build GRCh38
rs778349954 Chr.16: 58503978-58503978 on Build GRCh38
rs753521316 Chr.16: 58503963-58503964 on Build GRCh38
rs747673873 Chr.16: 58503984-58503984 on Build GRCh38
rs200016245 Chr.16: 58503821-58503821 on Build GRCh38
rs755853915 Chr.16: 58503974-58503974 on Build GRCh38
rs932983240 Chr.16: 58503849-58503849 on Build GRCh38
rs142554562 Chr.16: 58503852-58503852 on Build GRCh38
rs368353656 Chr.16: 58503845-58503845 on Build GRCh38
rs749311121 Chr.16: 58503934-58503934 on Build GRCh38
rs144560470 Chr.16: 58503809-58503809 on Build GRCh38
rs753890974 Chr.16: 58503900-58503900 on Build GRCh38
rs762624308 Chr.16: 58503806-58503806 on Build GRCh38
rs752854778 Chr.16: 58503969-58503969 on Build GRCh38
rs773570661 Chr.16: 58503954-58503954 on Build GRCh38
rs549958892 Chr.16: 58503921-58503921 on Build GRCh38
rs781094472 Chr.16: 58503914-58503914 on Build GRCh38
rs760954475 Chr.16: 58503956-58503956 on Build GRCh38
rs766835217 Chr.16: 58503957-58503957 on Build GRCh38
rs778464144 Chr.16: 58503818-58503818 on Build GRCh38
rs754287347 Chr.16: 58503965-58503965 on Build GRCh38
rs72790250 Chr.16: 58503873-58503873 on Build GRCh38
rs756623435 Chr.16: 58503820-58503820 on Build GRCh38
rs773985953 Chr.16: 58503868-58503868 on Build GRCh38

View Full Product Details