Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21179384-21179641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM159
Entrez Gene ID
57146
Gene Name
transmembrane protein 159
Gene Aliases
-
UniGene
Hs.258212
Gene Chromosome Location
Chr.16: 21158349-21180616 on Build GRCh38
Transcripts
NM_001301769.1 NM_001301771.1 NM_001301773.1 NM_001301774.1 NM_001301775.1 NM_020422.5 NR_125971.1 XM_005255439.4 XM_006721066.2 XM_006721069.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6990225 c.452A>T p.K151M Chr.16: 21179522-21179522 on Build GRCh38
6803450 c.445G>A p.A149T Chr.16: 21179515-21179515 on Build GRCh38
6803449 c.467A>G p.E156G Chr.16: 21179537-21179537 on Build GRCh38
81826 c.465C>T p.F155F Chr.16: 21179535-21179535 on Build GRCh38
5054729 c.442C>T p.P148S Chr.16: 21179512-21179512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1063087 Chr.16: 21179532-21179532 on Build GRCh38
rs368398652 Chr.16: 21179422-21179422 on Build GRCh38
rs758585149 Chr.16: 21179455-21179455 on Build GRCh38
rs752874647 Chr.16: 21179451-21179451 on Build GRCh38
rs760988365 Chr.16: 21179584-21179584 on Build GRCh38
rs755870271 Chr.16: 21179559-21179559 on Build GRCh38
rs749973113 Chr.16: 21179490-21179490 on Build GRCh38
rs769802540 Chr.16: 21179557-21179557 on Build GRCh38
rs547210794 Chr.16: 21179611-21179611 on Build GRCh38
rs930898658 Chr.16: 21179563-21179563 on Build GRCh38
rs761815395 Chr.16: 21179452-21179452 on Build GRCh38
rs189278970 Chr.16: 21179573-21179573 on Build GRCh38
rs747989064 Chr.16: 21179538-21179538 on Build GRCh38
rs894820389 Chr.16: 21179604-21179604 on Build GRCh38
rs777081625 Chr.16: 21179589-21179589 on Build GRCh38
rs150731603 Chr.16: 21179536-21179536 on Build GRCh38
rs757802375 Chr.16: 21179485-21179485 on Build GRCh38
rs768566290 Chr.16: 21179441-21179441 on Build GRCh38
rs540468585 Chr.16: 21179477-21179477 on Build GRCh38
rs746854342 Chr.16: 21179427-21179427 on Build GRCh38
rs143013789 Chr.16: 21179470-21179470 on Build GRCh38
rs765321653 Chr.16: 21179420-21179420 on Build GRCh38
rs770860698 Chr.16: 21179454-21179454 on Build GRCh38
rs761182478 Chr.16: 21179473-21179473 on Build GRCh38
rs764314452 Chr.16: 21179586-21179586 on Build GRCh38
rs765821702 Chr.16: 21179608-21179608 on Build GRCh38
rs370882955 Chr.16: 21179463-21179463 on Build GRCh38
rs774267278 Chr.16: 21179558-21179558 on Build GRCh38
rs754238079 Chr.16: 21179482-21179482 on Build GRCh38
rs532980501 Chr.16: 21179530-21179530 on Build GRCh38
rs758023545 Chr.16: 21179502-21179502 on Build GRCh38
rs939874083 Chr.16: 21179535-21179535 on Build GRCh38
rs200657852 Chr.16: 21179574-21179574 on Build GRCh38
rs138423091 Chr.16: 21179469-21179469 on Build GRCh38
rs762192038 Chr.16: 21179603-21179603 on Build GRCh38
rs778242307 Chr.16: 21179486-21179486 on Build GRCh38
rs552400284 Chr.16: 21179604-21179605 on Build GRCh38
rs759792772 Chr.16: 21179465-21179465 on Build GRCh38

View Full Product Details