Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37495744-37496011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEAF6
Entrez Gene ID
64769
Gene Name
MYST/Esa1 associated factor 6
Gene Aliases
C1orf149, CENP-28, EAF6, NY-SAR-91
UniGene
Hs.17118
Gene Chromosome Location
Chr.1: 37489960-37514819 on Build GRCh38
Transcripts
NM_001270875.1 NM_001270876.1 NM_022756.5 NR_073090.1 NR_073091.1 NR_073092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007568 c.564-1G>A p.? Chr.1: 37495919-37495919 on Build GRCh38
5494621 c.501+7A>G p.? Chr.1: 37495878-37495878 on Build GRCh38
5494620 c.597+7A>G p.? Chr.1: 37495878-37495878 on Build GRCh38
4007569 c.468-1G>A p.? Chr.1: 37495919-37495919 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917355433 Chr.1: 37495768-37495768 on Build GRCh38
rs984530277 Chr.1: 37495889-37495889 on Build GRCh38
rs971712113 Chr.1: 37495862-37495862 on Build GRCh38
rs537079193 Chr.1: 37495822-37495822 on Build GRCh38
rs763284092 Chr.1: 37495956-37495956 on Build GRCh38
rs201133071 Chr.1: 37495872-37495872 on Build GRCh38
rs186990548 Chr.1: 37495803-37495803 on Build GRCh38
rs753433816 Chr.1: 37495838-37495838 on Build GRCh38
rs778415355 Chr.1: 37495851-37495851 on Build GRCh38
rs766709984 Chr.1: 37495916-37495916 on Build GRCh38
rs561384468 Chr.1: 37495773-37495773 on Build GRCh38
rs773417627 Chr.1: 37495919-37495919 on Build GRCh38
rs937820392 Chr.1: 37495945-37495945 on Build GRCh38
rs779691022 Chr.1: 37495876-37495876 on Build GRCh38
rs558517098 Chr.1: 37495837-37495837 on Build GRCh38
rs201681275 Chr.1: 37495871-37495871 on Build GRCh38
rs746878409 Chr.1: 37495895-37495895 on Build GRCh38
rs780983244 Chr.1: 37495857-37495858 on Build GRCh38
rs369361938 Chr.1: 37495959-37495959 on Build GRCh38
rs199928363 Chr.1: 37495893-37495893 on Build GRCh38
rs946065473 Chr.1: 37495794-37495794 on Build GRCh38
rs1056824560 Chr.1: 37495965-37495965 on Build GRCh38
rs373187423 Chr.1: 37495848-37495848 on Build GRCh38
rs371056354 Chr.1: 37495834-37495834 on Build GRCh38

View Full Product Details