Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161119287-161119530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001185092.1 NM_001185093.1 NM_001185094.1 NM_005600.2 XM_005245214.2 XM_005245215.4 XM_005245216.3 XM_006711341.3 XM_017001371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232690 c.353+5C>G p.? Chr.1: 161119393-161119393 on Build GRCh38
2086683 c.251T>C p.L84P Chr.1: 161119331-161119331 on Build GRCh38
6232691 c.308+5C>G p.? Chr.1: 161119393-161119393 on Build GRCh38
6280981 c.353+6A>G p.? Chr.1: 161119394-161119394 on Build GRCh38
4538607 c.211G>C p.E71Q Chr.1: 161119291-161119291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777019181 Chr.1: 161119422-161119422 on Build GRCh38
rs182544592 Chr.1: 161119498-161119498 on Build GRCh38
rs765536780 Chr.1: 161119459-161119459 on Build GRCh38
rs760180629 Chr.1: 161119482-161119483 on Build GRCh38
rs757827244 Chr.1: 161119352-161119352 on Build GRCh38
rs865999622 Chr.1: 161119454-161119454 on Build GRCh38
rs368551610 Chr.1: 161119421-161119421 on Build GRCh38
rs747923881 Chr.1: 161119403-161119403 on Build GRCh38
rs374488499 Chr.1: 161119420-161119420 on Build GRCh38
rs755195924 Chr.1: 161119493-161119493 on Build GRCh38
rs1015847680 Chr.1: 161119359-161119359 on Build GRCh38
rs781542577 Chr.1: 161119496-161119496 on Build GRCh38
rs756008243 Chr.1: 161119502-161119502 on Build GRCh38
rs749411208 Chr.1: 161119361-161119362 on Build GRCh38
rs374120135 Chr.1: 161119503-161119503 on Build GRCh38
rs754186210 Chr.1: 161119340-161119340 on Build GRCh38
rs770105258 Chr.1: 161119354-161119355 on Build GRCh38
rs780139091 Chr.1: 161119392-161119392 on Build GRCh38
rs1003934925 Chr.1: 161119356-161119356 on Build GRCh38
rs373270446 Chr.1: 161119328-161119328 on Build GRCh38
rs370194915 Chr.1: 161119472-161119472 on Build GRCh38
rs1028184351 Chr.1: 161119432-161119432 on Build GRCh38
rs749199127 Chr.1: 161119506-161119506 on Build GRCh38
rs769906170 Chr.1: 161119404-161119404 on Build GRCh38
rs765864873 Chr.1: 161119317-161119317 on Build GRCh38
rs530645398 Chr.1: 161119470-161119470 on Build GRCh38
rs747198080 Chr.1: 161119393-161119393 on Build GRCh38
rs753514511 Chr.1: 161119379-161119379 on Build GRCh38
rs757151544 Chr.1: 161119511-161119511 on Build GRCh38
rs1032373493 Chr.1: 161119483-161119483 on Build GRCh38
rs750547483 Chr.1: 161119374-161119374 on Build GRCh38
rs202107253 Chr.1: 161119477-161119477 on Build GRCh38
rs781203445 Chr.1: 161119402-161119402 on Build GRCh38
rs765702221 Chr.1: 161119365-161119365 on Build GRCh38
rs762138337 Chr.1: 161119431-161119431 on Build GRCh38
rs767909234 Chr.1: 161119501-161119501 on Build GRCh38
rs1003161207 Chr.1: 161119384-161119384 on Build GRCh38
rs149045755 Chr.1: 161119313-161119313 on Build GRCh38
rs755217151 Chr.1: 161119397-161119397 on Build GRCh38
rs773259076 Chr.1: 161119356-161119357 on Build GRCh38
rs377319317 Chr.1: 161119413-161119413 on Build GRCh38
rs775016660 Chr.1: 161119480-161119484 on Build GRCh38
rs564939993 Chr.1: 161119467-161119467 on Build GRCh38
rs151133485 Chr.1: 161119329-161119329 on Build GRCh38
rs200278216 Chr.1: 161119312-161119312 on Build GRCh38
rs764377826 Chr.1: 161119333-161119333 on Build GRCh38
rs371117752 Chr.1: 161119419-161119419 on Build GRCh38
rs371279306 Chr.1: 161119381-161119381 on Build GRCh38
rs775969042 Chr.1: 161119506-161119507 on Build GRCh38
rs1035507992 Chr.1: 161119395-161119395 on Build GRCh38
rs771070623 Chr.1: 161119361-161119361 on Build GRCh38
rs751656514 Chr.1: 161119488-161119488 on Build GRCh38
rs144610510 Chr.1: 161119411-161119411 on Build GRCh38
rs758425902 Chr.1: 161119383-161119383 on Build GRCh38
rs766380610 Chr.1: 161119480-161119480 on Build GRCh38

View Full Product Details