Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41745678-41745939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIPK4
Entrez Gene ID
54101
Gene Name
receptor interacting serine/threonine kinase 4
Gene Aliases
ANKK2, ANKRD3, DIK, NKRD3, PKK, PPS2, RIP4
UniGene
Hs.517310
Gene Chromosome Location
Chr.21: 41739369-41767089 on Build GRCh38
Transcripts
NM_020639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1578194 c.1018G>T p.D340Y Chr.21: 41745821-41745821 on Build GRCh38
5432352 c.833-1G>A p.? Chr.21: 41745863-41745863 on Build GRCh38
6526285 c.1051G>T p.V351L Chr.21: 41745788-41745788 on Build GRCh38
5432353 c.976G>A p.E326K Chr.21: 41745863-41745863 on Build GRCh38
6571596 c.988G>A p.E330K Chr.21: 41745851-41745851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772803033 Chr.21: 41745914-41745914 on Build GRCh38
rs755951528 Chr.21: 41745775-41745775 on Build GRCh38
rs140139203 Chr.21: 41745780-41745780 on Build GRCh38
rs532143018 Chr.21: 41745916-41745916 on Build GRCh38
rs759917298 Chr.21: 41745908-41745908 on Build GRCh38
rs367750820 Chr.21: 41745716-41745716 on Build GRCh38
rs761353391 Chr.21: 41745910-41745910 on Build GRCh38
rs769711949 Chr.21: 41745712-41745712 on Build GRCh38
rs766964767 Chr.21: 41745912-41745912 on Build GRCh38
rs755863486 Chr.21: 41745861-41745861 on Build GRCh38
rs372754087 Chr.21: 41745818-41745818 on Build GRCh38
rs753718769 Chr.21: 41745870-41745870 on Build GRCh38
rs751073625 Chr.21: 41745826-41745826 on Build GRCh38
rs575160614 Chr.21: 41745705-41745705 on Build GRCh38
rs761101042 Chr.21: 41745801-41745801 on Build GRCh38
rs766983850 Chr.21: 41745738-41745738 on Build GRCh38
rs753626024 Chr.21: 41745919-41745919 on Build GRCh38
rs370055721 Chr.21: 41745736-41745736 on Build GRCh38
rs200625686 Chr.21: 41745721-41745721 on Build GRCh38
rs149641623 Chr.21: 41745762-41745762 on Build GRCh38
rs199835696 Chr.21: 41745845-41745845 on Build GRCh38
rs563233846 Chr.21: 41745876-41745876 on Build GRCh38
rs754449278 Chr.21: 41745740-41745740 on Build GRCh38
rs374420592 Chr.21: 41745884-41745884 on Build GRCh38
rs774395767 Chr.21: 41745722-41745722 on Build GRCh38
rs142027135 Chr.21: 41745789-41745789 on Build GRCh38
rs763441785 Chr.21: 41745819-41745819 on Build GRCh38
rs746595711 Chr.21: 41745774-41745775 on Build GRCh38
rs773630226 Chr.21: 41745735-41745735 on Build GRCh38
rs548177359 Chr.21: 41745892-41745892 on Build GRCh38
rs761188892 Chr.21: 41745737-41745737 on Build GRCh38
rs771467006 Chr.21: 41745815-41745815 on Build GRCh38
rs200747405 Chr.21: 41745830-41745830 on Build GRCh38
rs144324321 Chr.21: 41745774-41745774 on Build GRCh38
rs766183549 Chr.21: 41745869-41745869 on Build GRCh38
rs368491376 Chr.21: 41745776-41745776 on Build GRCh38
rs773365822 Chr.21: 41745799-41745799 on Build GRCh38
rs540850130 Chr.21: 41745729-41745729 on Build GRCh38
rs779064424 Chr.21: 41745788-41745788 on Build GRCh38
rs199827829 Chr.21: 41745753-41745753 on Build GRCh38
rs762794283 Chr.21: 41745882-41745882 on Build GRCh38
rs747270065 Chr.21: 41745888-41745888 on Build GRCh38
rs975935579 Chr.21: 41745706-41745706 on Build GRCh38
rs554261668 Chr.21: 41745839-41745839 on Build GRCh38
rs552178423 Chr.21: 41745918-41745918 on Build GRCh38
rs750114704 Chr.21: 41745852-41745852 on Build GRCh38
rs1044539325 Chr.21: 41745905-41745905 on Build GRCh38
rs763732233 Chr.21: 41745754-41745754 on Build GRCh38
rs772335369 Chr.21: 41745797-41745797 on Build GRCh38
rs371810697 Chr.21: 41745909-41745909 on Build GRCh38
rs895527097 Chr.21: 41745833-41745833 on Build GRCh38
rs756986161 Chr.21: 41745807-41745808 on Build GRCh38
rs746297192 Chr.21: 41745903-41745903 on Build GRCh38
rs950498181 Chr.21: 41745917-41745917 on Build GRCh38
rs778785773 Chr.21: 41745878-41745878 on Build GRCh38
rs778028069 Chr.21: 41745885-41745885 on Build GRCh38
rs748247665 Chr.21: 41745880-41745880 on Build GRCh38
rs924545174 Chr.21: 41745777-41745777 on Build GRCh38
rs1033495503 Chr.21: 41745899-41745899 on Build GRCh38
rs751112640 Chr.21: 41745756-41745756 on Build GRCh38
rs756936645 Chr.21: 41745761-41745761 on Build GRCh38
rs368349532 Chr.21: 41745893-41745893 on Build GRCh38

View Full Product Details