Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48832197-48832428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCSK1N
Entrez Gene ID
27344
Gene Name
proprotein convertase subtilisin/kexin type 1 inhibitor
Gene Aliases
BigLEN, PEN, PROSAAS, SAAS, SCG8, SgVIII
UniGene
Hs.522640
Gene Chromosome Location
Chr.X: 48831092-48835638 on Build GRCh38
Transcripts
NM_013271.4

SNP Detail

SNP ID Chromosome Location
rs782041087 Chr.X: 48832226-48832226 on Build GRCh38
rs1021077441 Chr.X: 48832365-48832365 on Build GRCh38
rs782785986 Chr.X: 48832243-48832243 on Build GRCh38
rs781862002 Chr.X: 48832245-48832245 on Build GRCh38
rs782656548 Chr.X: 48832330-48832330 on Build GRCh38
rs11538178 Chr.X: 48832339-48832339 on Build GRCh38
rs1046300820 Chr.X: 48832268-48832268 on Build GRCh38
rs782469472 Chr.X: 48832358-48832359 on Build GRCh38
rs782409475 Chr.X: 48832401-48832402 on Build GRCh38
rs781808466 Chr.X: 48832301-48832301 on Build GRCh38
rs782491051 Chr.X: 48832248-48832248 on Build GRCh38
rs782562370 Chr.X: 48832323-48832323 on Build GRCh38
rs782744723 Chr.X: 48832287-48832287 on Build GRCh38
rs782110662 Chr.X: 48832363-48832363 on Build GRCh38
rs781944303 Chr.X: 48832214-48832214 on Build GRCh38
rs1037402915 Chr.X: 48832227-48832227 on Build GRCh38
rs11538180 Chr.X: 48832296-48832296 on Build GRCh38
rs376988691 Chr.X: 48832334-48832334 on Build GRCh38
rs782155825 Chr.X: 48832236-48832236 on Build GRCh38
rs1002013414 Chr.X: 48832289-48832289 on Build GRCh38
rs782059582 Chr.X: 48832219-48832219 on Build GRCh38
rs782261596 Chr.X: 48832335-48832335 on Build GRCh38
rs993856254 Chr.X: 48832246-48832246 on Build GRCh38
rs782696353 Chr.X: 48832223-48832223 on Build GRCh38

View Full Product Details