Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127744888-127745161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D3C
Entrez Gene ID
10044
Gene Name
SH2 domain containing 3C
Gene Aliases
CHAT, NSP3, PRO34088, SHEP1
UniGene
Hs.306412
Gene Chromosome Location
Chr.9: 127738317-127778769 on Build GRCh38
Transcripts
NM_001142531.1 NM_001142532.1 NM_001142533.1 NM_001142534.1 NM_001252334.1 NM_005489.3 NM_170600.2 XM_005251639.1 XM_011518114.1 XM_011518115.1 XM_011518117.2 XM_017014174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1191856 c.1319C>T p.S440F Chr.9: 127745045-127745045 on Build GRCh38
3325966 c.1369C>A p.P457T Chr.9: 127744995-127744995 on Build GRCh38
4463499 c.1294C>T p.P432S Chr.9: 127745070-127745070 on Build GRCh38
4725563 c.799C>T p.R267C Chr.9: 127745094-127745094 on Build GRCh38
5410441 c.1174C>T p.H392Y Chr.9: 127744986-127744986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766536861 Chr.9: 127744972-127744974 on Build GRCh38
rs961360516 Chr.9: 127745130-127745130 on Build GRCh38
rs138786524 Chr.9: 127744960-127744960 on Build GRCh38
rs2233510 Chr.9: 127745004-127745004 on Build GRCh38
rs150113099 Chr.9: 127745126-127745126 on Build GRCh38
rs781179615 Chr.9: 127745094-127745094 on Build GRCh38
rs559861860 Chr.9: 127744925-127744925 on Build GRCh38
rs746092833 Chr.9: 127745100-127745100 on Build GRCh38
rs781436419 Chr.9: 127745005-127745005 on Build GRCh38
rs746117510 Chr.9: 127745013-127745013 on Build GRCh38
rs778726095 Chr.9: 127744976-127744976 on Build GRCh38
rs774886876 Chr.9: 127745113-127745113 on Build GRCh38
rs762697130 Chr.9: 127745131-127745131 on Build GRCh38
rs199821667 Chr.9: 127744924-127744924 on Build GRCh38
rs765844909 Chr.9: 127745053-127745053 on Build GRCh38
rs758394286 Chr.9: 127744984-127744984 on Build GRCh38
rs758201513 Chr.9: 127745080-127745080 on Build GRCh38
rs766004467 Chr.9: 127744944-127744944 on Build GRCh38
rs759898004 Chr.9: 127744911-127744911 on Build GRCh38
rs2233509 Chr.9: 127745055-127745055 on Build GRCh38
rs763607740 Chr.9: 127744915-127744915 on Build GRCh38
rs201064494 Chr.9: 127745119-127745119 on Build GRCh38
rs753130995 Chr.9: 127745126-127745127 on Build GRCh38
rs747054393 Chr.9: 127744996-127744996 on Build GRCh38
rs368075724 Chr.9: 127745034-127745034 on Build GRCh38
rs753462277 Chr.9: 127744957-127744957 on Build GRCh38
rs753945670 Chr.9: 127745007-127745012 on Build GRCh38
rs201761445 Chr.9: 127744978-127744978 on Build GRCh38
rs371293895 Chr.9: 127745127-127745127 on Build GRCh38
rs2233507 Chr.9: 127745111-127745111 on Build GRCh38
rs1045947488 Chr.9: 127745049-127745050 on Build GRCh38
rs770086336 Chr.9: 127744912-127744912 on Build GRCh38
rs776057887 Chr.9: 127745030-127745030 on Build GRCh38
rs2233508 Chr.9: 127745109-127745109 on Build GRCh38
rs151296579 Chr.9: 127745038-127745038 on Build GRCh38
rs776110834 Chr.9: 127744914-127744914 on Build GRCh38
rs777690834 Chr.9: 127744985-127744985 on Build GRCh38
rs987005618 Chr.9: 127744949-127744949 on Build GRCh38
rs765794901 Chr.9: 127745123-127745124 on Build GRCh38
rs1016330774 Chr.9: 127745079-127745079 on Build GRCh38
rs770458749 Chr.9: 127745117-127745117 on Build GRCh38
rs948789968 Chr.9: 127745031-127745031 on Build GRCh38
rs61761896 Chr.9: 127745037-127745037 on Build GRCh38
rs534443657 Chr.9: 127745057-127745057 on Build GRCh38
rs978473227 Chr.9: 127745003-127745003 on Build GRCh38
rs1028428206 Chr.9: 127744929-127744929 on Build GRCh38
rs560290474 Chr.9: 127745093-127745093 on Build GRCh38
rs751494340 Chr.9: 127745142-127745142 on Build GRCh38
rs756473394 Chr.9: 127745102-127745102 on Build GRCh38
rs770176536 Chr.9: 127745026-127745026 on Build GRCh38
rs777457171 Chr.9: 127745019-127745019 on Build GRCh38
rs760111430 Chr.9: 127744932-127744932 on Build GRCh38
rs758729318 Chr.9: 127745127-127745128 on Build GRCh38
rs548003990 Chr.9: 127745083-127745083 on Build GRCh38
rs763971193 Chr.9: 127745082-127745082 on Build GRCh38
rs752422850 Chr.9: 127745061-127745061 on Build GRCh38
rs760156196 Chr.9: 127745111-127745112 on Build GRCh38
rs775146341 Chr.9: 127745128-127745128 on Build GRCh38
rs763919760 Chr.9: 127745133-127745133 on Build GRCh38

View Full Product Details