Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27439737-27439968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRBP1
Entrez Gene ID
29959
Gene Name
nuclear receptor binding protein 1
Gene Aliases
BCON3, MADM, MUDPNP, NRBP
UniGene
Hs.515876
Gene Chromosome Location
Chr.2: 27427790-27442259 on Build GRCh38
Transcripts
NM_001321357.1 NM_001321358.1 NM_001321359.1 NM_001321361.1 NM_001321362.1 NM_001321363.1 NM_013392.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7175793 c.995C>T p.S332L Chr.2: 27439857-27439857 on Build GRCh38
5058855 c.932A>C p.E311A Chr.2: 27439794-27439794 on Build GRCh38
1407456 c.944G>A p.R315K Chr.2: 27439806-27439806 on Build GRCh38
78199 c.976G>C p.A326P Chr.2: 27439838-27439838 on Build GRCh38
6327719 c.958G>A p.E320K Chr.2: 27439820-27439820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764631733 Chr.2: 27439821-27439821 on Build GRCh38
rs145354088 Chr.2: 27439884-27439884 on Build GRCh38
rs764542054 Chr.2: 27439934-27439934 on Build GRCh38
rs965599439 Chr.2: 27439945-27439945 on Build GRCh38
rs772672860 Chr.2: 27439898-27439898 on Build GRCh38
rs536923313 Chr.2: 27439802-27439802 on Build GRCh38
rs750674505 Chr.2: 27439859-27439859 on Build GRCh38
rs758065640 Chr.2: 27439872-27439872 on Build GRCh38
rs764953490 Chr.2: 27439786-27439786 on Build GRCh38
rs1047100265 Chr.2: 27439780-27439780 on Build GRCh38
rs982142158 Chr.2: 27439816-27439816 on Build GRCh38
rs374669922 Chr.2: 27439913-27439913 on Build GRCh38
rs781321441 Chr.2: 27439885-27439885 on Build GRCh38
rs762662795 Chr.2: 27439905-27439905 on Build GRCh38
rs756627105 Chr.2: 27439813-27439813 on Build GRCh38
rs763695428 Chr.2: 27439919-27439920 on Build GRCh38
rs371384227 Chr.2: 27439892-27439892 on Build GRCh38
rs576571594 Chr.2: 27439937-27439937 on Build GRCh38
rs71441077 Chr.2: 27439826-27439826 on Build GRCh38
rs367815724 Chr.2: 27439873-27439873 on Build GRCh38
rs201132320 Chr.2: 27439800-27439800 on Build GRCh38
rs201596708 Chr.2: 27439841-27439841 on Build GRCh38
rs141001145 Chr.2: 27439897-27439897 on Build GRCh38
rs770543951 Chr.2: 27439906-27439906 on Build GRCh38
rs371237665 Chr.2: 27439803-27439803 on Build GRCh38
rs746838386 Chr.2: 27439876-27439876 on Build GRCh38
rs71441076 Chr.2: 27439791-27439791 on Build GRCh38
rs773019385 Chr.2: 27439795-27439795 on Build GRCh38
rs375471593 Chr.2: 27439858-27439858 on Build GRCh38
rs760982004 Chr.2: 27439922-27439922 on Build GRCh38
rs757360840 Chr.2: 27439828-27439828 on Build GRCh38

View Full Product Details