Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204954065-204954326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFASC
Entrez Gene ID
23114
Gene Name
neurofascin
Gene Aliases
NF, NRCAML
UniGene
Hs.13349
Gene Chromosome Location
Chr.1: 204828652-205022822 on Build GRCh38
Transcripts
NM_001005388.2 NM_001005389.1 NM_001160331.1 NM_001160332.1 NM_001160333.1 NM_015090.3 XM_005244989.3 XM_005244991.3 XM_005244992.4 XM_005244993.3 XM_005244997.3 XM_011509311.1 XM_011509312.1 XM_011509313.1 XM_011509314.1 XM_011509315.1 XM_011509316.1 XM_011509317.1 XM_011509318.2 XM_011509319.1 XM_011509320.2 XM_011509321.1 XM_011509322.1 XM_011509323.1 XM_011509324.1 XM_011509325.1 XM_011509326.1 XM_011509327.1 XM_011509328.1 XM_017000733.1 XM_017000734.1 XM_017000735.1 XM_017000736.1 XM_017000737.1 XM_017000738.1 XM_017000739.1 XM_017000740.1 XM_017000741.1 XM_017000742.1 XM_017000743.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79396 c.206G>A p.W69* Chr.1: 204954196-204954196 on Build GRCh38
4229597 c.246C>G p.F82L Chr.1: 204954218-204954218 on Build GRCh38
4964361 c.300C>T p.G100G Chr.1: 204954290-204954290 on Build GRCh38
6244445 c.300G>T p.V100V Chr.1: 204954272-204954272 on Build GRCh38
5353334 c.323G>A p.R108Q Chr.1: 204954295-204954295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971729346 Chr.1: 204954194-204954194 on Build GRCh38
rs769904507 Chr.1: 204954225-204954225 on Build GRCh38
rs951157942 Chr.1: 204954093-204954093 on Build GRCh38
rs564524486 Chr.1: 204954295-204954295 on Build GRCh38
rs777346580 Chr.1: 204954164-204954164 on Build GRCh38
rs375752918 Chr.1: 204954224-204954224 on Build GRCh38
rs778316321 Chr.1: 204954286-204954286 on Build GRCh38
rs144793383 Chr.1: 204954238-204954238 on Build GRCh38
rs772181654 Chr.1: 204954248-204954248 on Build GRCh38
rs775540551 Chr.1: 204954231-204954231 on Build GRCh38
rs552380308 Chr.1: 204954129-204954129 on Build GRCh38
rs780974163 Chr.1: 204954297-204954297 on Build GRCh38
rs142536240 Chr.1: 204954290-204954290 on Build GRCh38
rs147387188 Chr.1: 204954278-204954278 on Build GRCh38
rs781090491 Chr.1: 204954216-204954216 on Build GRCh38
rs746612240 Chr.1: 204954185-204954185 on Build GRCh38
rs773282718 Chr.1: 204954288-204954291 on Build GRCh38
rs756631665 Chr.1: 204954198-204954198 on Build GRCh38
rs757920700 Chr.1: 204954155-204954155 on Build GRCh38
rs187989406 Chr.1: 204954115-204954115 on Build GRCh38
rs753232079 Chr.1: 204954285-204954285 on Build GRCh38
rs763590330 Chr.1: 204954291-204954291 on Build GRCh38
rs149736553 Chr.1: 204954298-204954298 on Build GRCh38
rs139082842 Chr.1: 204954263-204954263 on Build GRCh38
rs145530574 Chr.1: 204954267-204954267 on Build GRCh38
rs1043139762 Chr.1: 204954245-204954245 on Build GRCh38
rs757003214 Chr.1: 204954202-204954202 on Build GRCh38
rs767296212 Chr.1: 204954282-204954282 on Build GRCh38
rs947517631 Chr.1: 204954235-204954235 on Build GRCh38
rs765710562 Chr.1: 204954145-204954145 on Build GRCh38
rs751198725 Chr.1: 204954294-204954294 on Build GRCh38
rs759862571 Chr.1: 204954283-204954283 on Build GRCh38
rs755438381 Chr.1: 204954143-204954143 on Build GRCh38
rs202239718 Chr.1: 204954299-204954299 on Build GRCh38
rs748244825 Chr.1: 204954239-204954239 on Build GRCh38
rs563452835 Chr.1: 204954108-204954108 on Build GRCh38
rs922818448 Chr.1: 204954237-204954237 on Build GRCh38
rs12068836 Chr.1: 204954154-204954154 on Build GRCh38
rs760885949 Chr.1: 204954270-204954270 on Build GRCh38

View Full Product Details