Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120212089-120212362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXN
Entrez Gene ID
5829
Gene Name
paxillin
Gene Aliases
-
UniGene
Hs.446336
Gene Chromosome Location
Chr.12: 120210439-120265771 on Build GRCh38
Transcripts
NM_001080855.2 NM_001243756.1 NM_002859.3 NM_025157.4 XM_005253917.3 XM_006719532.1 XM_006719534.1 XM_006719535.3 XM_006719536.2 XM_011538622.1 XM_011538623.1 XM_017019726.1 XM_017019727.1 XM_017019728.1 XM_017019729.1 XM_017019731.1 XM_017019733.1 XM_017019734.1 XM_017019735.1 XM_017019736.1 XM_017019737.1 XM_017019738.1 XM_017019739.1 XM_017019740.1 XM_017019741.1 XM_017019742.1 XM_017019743.1 NR_038924.1

Gene Symbol
PXN-AS1
Entrez Gene ID
100506649
Gene Name
PXN antisense RNA 1
Gene Aliases
EyeLinc4
UniGene
Hs.569205
Gene Chromosome Location
Chr.12: 120201291-120212828 on Build GRCh38
Transcripts
NM_001080855.2 NM_001243756.1 NM_002859.3 NM_025157.4 XM_005253917.3 XM_006719532.1 XM_006719534.1 XM_006719535.3 XM_006719536.2 XM_011538622.1 XM_011538623.1 XM_017019726.1 XM_017019727.1 XM_017019728.1 XM_017019729.1 XM_017019731.1 XM_017019733.1 XM_017019734.1 XM_017019735.1 XM_017019736.1 XM_017019737.1 XM_017019738.1 XM_017019739.1 XM_017019740.1 XM_017019741.1 XM_017019742.1 XM_017019743.1 NR_038924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7348724 c.1732G>A p.D578N Chr.12: 120212358-120212358 on Build GRCh38
7120195 c.1668C>G p.F556L Chr.12: 120212320-120212320 on Build GRCh38
7120196 c.1206C>G p.F402L Chr.12: 120212320-120212320 on Build GRCh38
4864346 c.1173G>A p.K391K Chr.12: 120212353-120212353 on Build GRCh38
6760686 c.1167C>T p.N389N Chr.12: 120212359-120212359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982094583 Chr.12: 120212203-120212203 on Build GRCh38
rs761311631 Chr.12: 120212289-120212290 on Build GRCh38
rs920726681 Chr.12: 120212263-120212263 on Build GRCh38
rs748911095 Chr.12: 120212286-120212286 on Build GRCh38
rs772832049 Chr.12: 120212301-120212301 on Build GRCh38
rs541907834 Chr.12: 120212192-120212192 on Build GRCh38
rs762172536 Chr.12: 120212293-120212293 on Build GRCh38
rs769477361 Chr.12: 120212267-120212267 on Build GRCh38
rs770644352 Chr.12: 120212193-120212193 on Build GRCh38
rs750611347 Chr.12: 120212305-120212305 on Build GRCh38
rs555376512 Chr.12: 120212218-120212218 on Build GRCh38
rs761152303 Chr.12: 120212290-120212290 on Build GRCh38
rs957987473 Chr.12: 120212145-120212145 on Build GRCh38
rs1016264697 Chr.12: 120212224-120212224 on Build GRCh38
rs769355352 Chr.12: 120212301-120212302 on Build GRCh38
rs1051157851 Chr.12: 120212307-120212307 on Build GRCh38
rs564545490 Chr.12: 120212235-120212235 on Build GRCh38
rs576940787 Chr.12: 120212151-120212151 on Build GRCh38
rs573194301 Chr.12: 120212180-120212180 on Build GRCh38
rs3742039 Chr.12: 120212167-120212167 on Build GRCh38
rs972238764 Chr.12: 120212239-120212239 on Build GRCh38
rs889369953 Chr.12: 120212308-120212308 on Build GRCh38
rs551071061 Chr.12: 120212234-120212234 on Build GRCh38
rs562117346 Chr.12: 120212204-120212204 on Build GRCh38
rs545967803 Chr.12: 120212148-120212148 on Build GRCh38
rs1052656746 Chr.12: 120212175-120212175 on Build GRCh38
rs765045352 Chr.12: 120212200-120212200 on Build GRCh38
rs773894457 Chr.12: 120212288-120212288 on Build GRCh38
rs777423522 Chr.12: 120212275-120212275 on Build GRCh38
rs765535652 Chr.12: 120212298-120212298 on Build GRCh38
rs769212427 Chr.12: 120212292-120212292 on Build GRCh38
rs889193993 Chr.12: 120212210-120212210 on Build GRCh38
rs924962171 Chr.12: 120212159-120212159 on Build GRCh38
rs942400610 Chr.12: 120212340-120212340 on Build GRCh38
rs577042317 Chr.12: 120212118-120212118 on Build GRCh38
rs140367844 Chr.12: 120212306-120212306 on Build GRCh38

View Full Product Details