Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21461123-21461372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB2B
Entrez Gene ID
84932
Gene Name
RAB2B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.22399
Gene Chromosome Location
Chr.14: 21459020-21476973 on Build GRCh38
Transcripts
NM_001163380.1 NM_032846.3 NR_028074.1 XM_017021710.1 XM_017021711.1 XM_017021712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846969 c.612G>A p.R204R Chr.14: 21461235-21461235 on Build GRCh38
5582292 c.605C>T p.S202F Chr.14: 21461242-21461242 on Build GRCh38
6232262 c.566C>T p.P189L Chr.14: 21461281-21461281 on Build GRCh38
5600333 c.606C>T p.S202S Chr.14: 21461241-21461241 on Build GRCh38
4807044 c.611G>A p.R204Q Chr.14: 21461236-21461236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765160261 Chr.14: 21461250-21461250 on Build GRCh38
rs761679847 Chr.14: 21461237-21461237 on Build GRCh38
rs751196214 Chr.14: 21461277-21461277 on Build GRCh38
rs933067206 Chr.14: 21461326-21461326 on Build GRCh38
rs762053253 Chr.14: 21461217-21461217 on Build GRCh38
rs145245060 Chr.14: 21461280-21461280 on Build GRCh38
rs770985562 Chr.14: 21461219-21461219 on Build GRCh38
rs772293443 Chr.14: 21461341-21461341 on Build GRCh38
rs779749833 Chr.14: 21461174-21461174 on Build GRCh38
rs140211092 Chr.14: 21461228-21461228 on Build GRCh38
rs773369232 Chr.14: 21461213-21461213 on Build GRCh38
rs750423406 Chr.14: 21461260-21461260 on Build GRCh38
rs371539903 Chr.14: 21461327-21461327 on Build GRCh38
rs764676477 Chr.14: 21461154-21461154 on Build GRCh38
rs757216332 Chr.14: 21461153-21461153 on Build GRCh38
rs765170882 Chr.14: 21461308-21461312 on Build GRCh38
rs960077345 Chr.14: 21461163-21461163 on Build GRCh38
rs765501616 Chr.14: 21461342-21461343 on Build GRCh38
rs774452404 Chr.14: 21461244-21461244 on Build GRCh38
rs376736457 Chr.14: 21461185-21461185 on Build GRCh38
rs771010618 Chr.14: 21461328-21461328 on Build GRCh38
rs984540817 Chr.14: 21461214-21461214 on Build GRCh38
rs756624337 Chr.14: 21461315-21461315 on Build GRCh38
rs774324252 Chr.14: 21461333-21461333 on Build GRCh38
rs781651824 Chr.14: 21461286-21461286 on Build GRCh38
rs746053735 Chr.14: 21461339-21461339 on Build GRCh38
rs61752840 Chr.14: 21461208-21461208 on Build GRCh38
rs1041182311 Chr.14: 21461252-21461252 on Build GRCh38
rs749391593 Chr.14: 21461216-21461216 on Build GRCh38
rs749946471 Chr.14: 21461166-21461166 on Build GRCh38
rs776432456 Chr.14: 21461236-21461236 on Build GRCh38
rs765853146 Chr.14: 21461275-21461275 on Build GRCh38
rs778421065 Chr.14: 21461320-21461320 on Build GRCh38
rs757918148 Chr.14: 21461170-21461170 on Build GRCh38
rs781720234 Chr.14: 21461191-21461191 on Build GRCh38
rs755350193 Chr.14: 21461188-21461188 on Build GRCh38
rs757910545 Chr.14: 21461263-21461263 on Build GRCh38
rs17106411 Chr.14: 21461212-21461212 on Build GRCh38
rs200862962 Chr.14: 21461311-21461311 on Build GRCh38
rs891284144 Chr.14: 21461222-21461222 on Build GRCh38

View Full Product Details