Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46899106-46899377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTH1R
Entrez Gene ID
5745
Gene Name
parathyroid hormone 1 receptor
Gene Aliases
PFE, PTHR, PTHR1
UniGene
Hs.1019
Gene Chromosome Location
Chr.3: 46877690-46903799 on Build GRCh38
Transcripts
NM_000316.2 NM_001184744.1 XM_005265344.3 XM_011533967.2 XM_011533968.2 XM_017006932.1 XM_017006933.1 XM_017006934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
269971 c.851C>T p.A284V Chr.3: 46899319-46899319 on Build GRCh38
1045354 c.865C>A p.L289I Chr.3: 46899333-46899333 on Build GRCh38
6329944 c.869A>C p.Y290S Chr.3: 46899337-46899337 on Build GRCh38
1423470 c.837G>A p.A279A Chr.3: 46899305-46899305 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs568989577 Chr.3: 46899207-46899207 on Build GRCh38
rs945757482 Chr.3: 46899309-46899309 on Build GRCh38
rs41290650 Chr.3: 46899260-46899260 on Build GRCh38
rs776293238 Chr.3: 46899268-46899268 on Build GRCh38
rs200475872 Chr.3: 46899344-46899344 on Build GRCh38
rs754223892 Chr.3: 46899157-46899157 on Build GRCh38
rs889678343 Chr.3: 46899227-46899228 on Build GRCh38
rs981377741 Chr.3: 46899204-46899204 on Build GRCh38
rs1041429425 Chr.3: 46899249-46899249 on Build GRCh38
rs769883740 Chr.3: 46899304-46899304 on Build GRCh38
rs954466379 Chr.3: 46899171-46899171 on Build GRCh38
rs387907459 Chr.3: 46899343-46899343 on Build GRCh38
rs944075101 Chr.3: 46899238-46899238 on Build GRCh38
rs555011284 Chr.3: 46899150-46899151 on Build GRCh38
rs753946111 Chr.3: 46899355-46899355 on Build GRCh38
rs934399522 Chr.3: 46899149-46899149 on Build GRCh38
rs779811103 Chr.3: 46899258-46899258 on Build GRCh38
rs202059785 Chr.3: 46899351-46899351 on Build GRCh38
rs914336245 Chr.3: 46899291-46899291 on Build GRCh38
rs748048870 Chr.3: 46899293-46899293 on Build GRCh38
rs1049679584 Chr.3: 46899208-46899208 on Build GRCh38
rs754222457 Chr.3: 46899308-46899308 on Build GRCh38
rs1016391519 Chr.3: 46899159-46899159 on Build GRCh38
rs371454798 Chr.3: 46899338-46899338 on Build GRCh38
rs750936657 Chr.3: 46899256-46899256 on Build GRCh38
rs971320996 Chr.3: 46899203-46899203 on Build GRCh38
rs962400893 Chr.3: 46899176-46899176 on Build GRCh38
rs201499957 Chr.3: 46899330-46899330 on Build GRCh38
rs1025364430 Chr.3: 46899194-46899194 on Build GRCh38
rs990386296 Chr.3: 46899283-46899283 on Build GRCh38
rs762855724 Chr.3: 46899318-46899318 on Build GRCh38
rs577419212 Chr.3: 46899190-46899190 on Build GRCh38
rs559583997 Chr.3: 46899326-46899326 on Build GRCh38
rs988642262 Chr.3: 46899156-46899156 on Build GRCh38
rs746494701 Chr.3: 46899259-46899259 on Build GRCh38
rs764106190 Chr.3: 46899350-46899350 on Build GRCh38
rs558964541 Chr.3: 46899164-46899164 on Build GRCh38
rs928429598 Chr.3: 46899147-46899147 on Build GRCh38

View Full Product Details