Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119673879-119674140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAB1
Entrez Gene ID
5564
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 1
Gene Aliases
AMPK, HAMPKb
UniGene
Hs.741184
Gene Chromosome Location
Chr.12: 119667864-119681624 on Build GRCh38
Transcripts
NM_006253.4 XM_005253909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6537842 c.333C>G p.N111K Chr.12: 119673973-119673973 on Build GRCh38
6755616 c.399G>T p.W133C Chr.12: 119674039-119674039 on Build GRCh38
3687987 c.405C>T p.H135H Chr.12: 119674045-119674045 on Build GRCh38
4408195 c.350A>G p.D117G Chr.12: 119673990-119673990 on Build GRCh38
1561814 c.417G>T p.E139D Chr.12: 119674057-119674057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747611543 Chr.12: 119674042-119674042 on Build GRCh38
rs781002487 Chr.12: 119674041-119674041 on Build GRCh38
rs757346466 Chr.12: 119673996-119673996 on Build GRCh38
rs1041116805 Chr.12: 119673930-119673930 on Build GRCh38
rs750394660 Chr.12: 119673998-119673998 on Build GRCh38
rs775919641 Chr.12: 119674065-119674065 on Build GRCh38
rs373505242 Chr.12: 119674046-119674046 on Build GRCh38
rs202243548 Chr.12: 119674017-119674017 on Build GRCh38
rs193278535 Chr.12: 119674050-119674050 on Build GRCh38
rs779535233 Chr.12: 119673921-119673921 on Build GRCh38
rs372075157 Chr.12: 119674079-119674079 on Build GRCh38
rs146404701 Chr.12: 119674054-119674054 on Build GRCh38
rs761408157 Chr.12: 119673938-119673938 on Build GRCh38
rs538915515 Chr.12: 119673988-119673988 on Build GRCh38
rs781402276 Chr.12: 119673914-119673915 on Build GRCh38
rs74516777 Chr.12: 119674023-119674023 on Build GRCh38
rs764289933 Chr.12: 119673991-119673991 on Build GRCh38
rs982072870 Chr.12: 119674084-119674084 on Build GRCh38
rs142228541 Chr.12: 119674045-119674045 on Build GRCh38
rs201378473 Chr.12: 119674094-119674094 on Build GRCh38
rs753560304 Chr.12: 119673946-119673949 on Build GRCh38
rs775020808 Chr.12: 119673957-119673957 on Build GRCh38
rs767890601 Chr.12: 119673977-119673977 on Build GRCh38
rs771593569 Chr.12: 119673974-119673975 on Build GRCh38
rs776274834 Chr.12: 119673934-119673934 on Build GRCh38
rs757038497 Chr.12: 119674053-119674053 on Build GRCh38
rs760939482 Chr.12: 119673990-119673990 on Build GRCh38
rs761843387 Chr.12: 119674106-119674106 on Build GRCh38
rs746645237 Chr.12: 119673931-119673931 on Build GRCh38
rs368579544 Chr.12: 119674055-119674055 on Build GRCh38
rs76843493 Chr.12: 119674093-119674093 on Build GRCh38
rs748078282 Chr.12: 119673915-119673915 on Build GRCh38
rs758310763 Chr.12: 119674004-119674004 on Build GRCh38
rs1008554041 Chr.12: 119673906-119673906 on Build GRCh38
rs374561238 Chr.12: 119673997-119673997 on Build GRCh38
rs759777721 Chr.12: 119673973-119673973 on Build GRCh38
rs970816835 Chr.12: 119674075-119674075 on Build GRCh38
rs1021020896 Chr.12: 119674072-119674072 on Build GRCh38
rs549700674 Chr.12: 119673914-119673914 on Build GRCh38
rs771928281 Chr.12: 119673917-119673917 on Build GRCh38
rs779882642 Chr.12: 119674006-119674006 on Build GRCh38
rs370057551 Chr.12: 119673933-119673933 on Build GRCh38
rs771505857 Chr.12: 119673953-119673953 on Build GRCh38
rs11546695 Chr.12: 119674044-119674044 on Build GRCh38

View Full Product Details