Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27032939-27033182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM214
Entrez Gene ID
54867
Gene Name
transmembrane protein 214
Gene Aliases
-
UniGene
Hs.533934
Gene Chromosome Location
Chr.2: 27032906-27041697 on Build GRCh38
Transcripts
NM_001083590.1 NM_017727.4 XM_005264381.3 XM_005264382.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6502283 c.100A>G p.N34D Chr.2: 27033115-27033115 on Build GRCh38
6865762 c.130G>T p.V44L Chr.2: 27033145-27033145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1039311893 Chr.2: 27033009-27033009 on Build GRCh38
rs758562755 Chr.2: 27033063-27033076 on Build GRCh38
rs945396615 Chr.2: 27033063-27033063 on Build GRCh38
rs766687480 Chr.2: 27032975-27032975 on Build GRCh38
rs902158995 Chr.2: 27033034-27033034 on Build GRCh38
rs1056658474 Chr.2: 27033025-27033025 on Build GRCh38
rs1000942485 Chr.2: 27033080-27033081 on Build GRCh38
rs1039103172 Chr.2: 27033084-27033084 on Build GRCh38
rs1032593644 Chr.2: 27033068-27033068 on Build GRCh38
rs751885270 Chr.2: 27033001-27033001 on Build GRCh38
rs368291134 Chr.2: 27033106-27033106 on Build GRCh38
rs999626850 Chr.2: 27033040-27033040 on Build GRCh38
rs958567151 Chr.2: 27033072-27033072 on Build GRCh38
rs574052269 Chr.2: 27032986-27032986 on Build GRCh38
rs949416504 Chr.2: 27032962-27032962 on Build GRCh38
rs991436326 Chr.2: 27032956-27032956 on Build GRCh38
rs546821657 Chr.2: 27033096-27033096 on Build GRCh38
rs997182372 Chr.2: 27033027-27033027 on Build GRCh38
rs866466271 Chr.2: 27032960-27032960 on Build GRCh38
rs916663747 Chr.2: 27032959-27032959 on Build GRCh38
rs887712464 Chr.2: 27032994-27032994 on Build GRCh38
rs997327025 Chr.2: 27033139-27033139 on Build GRCh38
rs766583742 Chr.2: 27033081-27033086 on Build GRCh38
rs982197448 Chr.2: 27032964-27032964 on Build GRCh38
rs866636317 Chr.2: 27033000-27033000 on Build GRCh38
rs372186280 Chr.2: 27033010-27033010 on Build GRCh38
rs1047629137 Chr.2: 27032979-27032979 on Build GRCh38
rs918569225 Chr.2: 27032969-27032969 on Build GRCh38
rs974253849 Chr.2: 27032985-27032985 on Build GRCh38
rs1012510088 Chr.2: 27033076-27033076 on Build GRCh38
rs867197514 Chr.2: 27033071-27033071 on Build GRCh38
rs941868725 Chr.2: 27033007-27033007 on Build GRCh38
rs375080415 Chr.2: 27033030-27033030 on Build GRCh38
rs752510848 Chr.2: 27033093-27033098 on Build GRCh38
rs578217345 Chr.2: 27033067-27033067 on Build GRCh38
rs1042372144 Chr.2: 27033069-27033069 on Build GRCh38

View Full Product Details