Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 198277892-198278147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK7
Entrez Gene ID
140609
Gene Name
NIMA related kinase 7
Gene Aliases
-
UniGene
Hs.24119
Gene Chromosome Location
Chr.1: 198156978-198322420 on Build GRCh38
Transcripts
NM_133494.2 XM_011509209.1 XM_017000344.1 XM_017000345.1 XM_017000346.1 XM_017000347.1 XM_017000348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6996689 c.456G>A p.M152I Chr.1: 198278044-198278044 on Build GRCh38
4026714 c.438C>T p.C146C Chr.1: 198278026-198278026 on Build GRCh38
5899831 c.373-7C>T p.? Chr.1: 198277954-198277954 on Build GRCh38
6724518 c.373-1G>A p.? Chr.1: 198277960-198277960 on Build GRCh38
6123790 c.432G>T p.Q144H Chr.1: 198278020-198278020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754449598 Chr.1: 198278048-198278048 on Build GRCh38
rs762809840 Chr.1: 198277974-198277974 on Build GRCh38
rs776687699 Chr.1: 198278091-198278091 on Build GRCh38
rs367944492 Chr.1: 198278027-198278027 on Build GRCh38
rs769982238 Chr.1: 198278097-198278097 on Build GRCh38
rs765447602 Chr.1: 198277929-198277929 on Build GRCh38
rs932932211 Chr.1: 198277941-198277941 on Build GRCh38
rs767283058 Chr.1: 198277945-198277945 on Build GRCh38
rs878985720 Chr.1: 198277947-198277947 on Build GRCh38
rs116369607 Chr.1: 198278099-198278099 on Build GRCh38
rs372494155 Chr.1: 198278043-198278043 on Build GRCh38
rs372111535 Chr.1: 198278088-198278088 on Build GRCh38
rs755751284 Chr.1: 198277983-198277983 on Build GRCh38
rs750187883 Chr.1: 198277961-198277961 on Build GRCh38
rs113240903 Chr.1: 198278081-198278081 on Build GRCh38
rs770357863 Chr.1: 198277942-198277942 on Build GRCh38
rs774428189 Chr.1: 198277935-198277935 on Build GRCh38
rs964825750 Chr.1: 198278000-198278000 on Build GRCh38
rs1049392766 Chr.1: 198278025-198278025 on Build GRCh38
rs765034929 Chr.1: 198278113-198278113 on Build GRCh38
rs931163651 Chr.1: 198278112-198278112 on Build GRCh38
rs778458670 Chr.1: 198278051-198278051 on Build GRCh38
rs191347826 Chr.1: 198278087-198278087 on Build GRCh38
rs748638300 Chr.1: 198278060-198278060 on Build GRCh38

View Full Product Details