Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101616221-101616458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL10
Entrez Gene ID
93408
Gene Name
myosin light chain 10
Gene Aliases
MYLC2PL, PLRLC
UniGene
Hs.247831
Gene Chromosome Location
Chr.7: 101613325-101629296 on Build GRCh38
Transcripts
NM_138403.4 XM_017012793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1083436 c.488T>G p.F163C Chr.7: 101616265-101616265 on Build GRCh38
291822 c.519C>T p.F173F Chr.7: 101616234-101616234 on Build GRCh38
4895414 c.468G>A p.E156E Chr.7: 101616285-101616285 on Build GRCh38
1719585 c.473C>T p.T158I Chr.7: 101616280-101616280 on Build GRCh38
4596142 c.509G>A p.G170E Chr.7: 101616244-101616244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767812700 Chr.7: 101616336-101616336 on Build GRCh38
rs920580986 Chr.7: 101616306-101616306 on Build GRCh38
rs757267402 Chr.7: 101616277-101616277 on Build GRCh38
rs755297614 Chr.7: 101616409-101616409 on Build GRCh38
rs746059992 Chr.7: 101616289-101616289 on Build GRCh38
rs779729708 Chr.7: 101616257-101616258 on Build GRCh38
rs753054867 Chr.7: 101616345-101616345 on Build GRCh38
rs761415917 Chr.7: 101616310-101616310 on Build GRCh38
rs756582817 Chr.7: 101616269-101616269 on Build GRCh38
rs577730144 Chr.7: 101616400-101616400 on Build GRCh38
rs776656052 Chr.7: 101616303-101616303 on Build GRCh38
rs368209720 Chr.7: 101616346-101616346 on Build GRCh38
rs142075052 Chr.7: 101616372-101616372 on Build GRCh38
rs762646362 Chr.7: 101616328-101616328 on Build GRCh38
rs777895650 Chr.7: 101616270-101616270 on Build GRCh38
rs750017494 Chr.7: 101616311-101616311 on Build GRCh38
rs201660825 Chr.7: 101616295-101616295 on Build GRCh38
rs907953866 Chr.7: 101616367-101616367 on Build GRCh38
rs947404064 Chr.7: 101616382-101616382 on Build GRCh38
rs986948028 Chr.7: 101616304-101616304 on Build GRCh38
rs537921580 Chr.7: 101616312-101616312 on Build GRCh38
rs764998096 Chr.7: 101616250-101616250 on Build GRCh38
rs35570198 Chr.7: 101616425-101616426 on Build GRCh38
rs779004791 Chr.7: 101616279-101616279 on Build GRCh38
rs767875981 Chr.7: 101616255-101616255 on Build GRCh38
rs895093441 Chr.7: 101616379-101616379 on Build GRCh38
rs950173330 Chr.7: 101616353-101616364 on Build GRCh38
rs1056294366 Chr.7: 101616377-101616377 on Build GRCh38
rs376056819 Chr.7: 101616291-101616291 on Build GRCh38
rs1044424613 Chr.7: 101616386-101616386 on Build GRCh38
rs567400190 Chr.7: 101616294-101616294 on Build GRCh38
rs930723542 Chr.7: 101616318-101616318 on Build GRCh38
rs754367637 Chr.7: 101616388-101616388 on Build GRCh38
rs548990601 Chr.7: 101616254-101616254 on Build GRCh38
rs749354639 Chr.7: 101616275-101616275 on Build GRCh38
rs372517822 Chr.7: 101616260-101616260 on Build GRCh38
rs772763019 Chr.7: 101616315-101616315 on Build GRCh38

View Full Product Details