Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69381848-69382101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD17
Entrez Gene ID
5884
Gene Name
RAD17 checkpoint clamp loader component
Gene Aliases
CCYC, HRAD17, R24L, RAD17SP, RAD24
UniGene
Hs.16184
Gene Chromosome Location
Chr.5: 69369297-69414801 on Build GRCh38
Transcripts
NM_001278622.1 NM_002873.1 NM_133338.2 NM_133339.2 NM_133340.2 NM_133341.2 NM_133342.2 NM_133343.1 NM_133344.2 XM_017009679.1 XM_017009680.1 XM_017009681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715262 c.408A>T p.L136F Chr.5: 69381957-69381957 on Build GRCh38
3920292 c.435G>A p.T145T Chr.5: 69381951-69381951 on Build GRCh38
4493352 c.380C>T p.P127L Chr.5: 69381929-69381929 on Build GRCh38
7386693 c.466A>T p.I156F Chr.5: 69381982-69381982 on Build GRCh38
4549461 c.439G>T p.V147L Chr.5: 69381988-69381988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761085514 Chr.5: 69382031-69382031 on Build GRCh38
rs377481140 Chr.5: 69382068-69382068 on Build GRCh38
rs751789713 Chr.5: 69382069-69382069 on Build GRCh38
rs762979455 Chr.5: 69382060-69382060 on Build GRCh38
rs955274008 Chr.5: 69382062-69382062 on Build GRCh38
rs752797808 Chr.5: 69382058-69382058 on Build GRCh38
rs767107512 Chr.5: 69381911-69381911 on Build GRCh38
rs779957091 Chr.5: 69381979-69381979 on Build GRCh38
rs760080251 Chr.5: 69382038-69382038 on Build GRCh38
rs777902658 Chr.5: 69381969-69381969 on Build GRCh38
rs776702424 Chr.5: 69381904-69381904 on Build GRCh38
rs760995950 Chr.5: 69381916-69381916 on Build GRCh38
rs749169139 Chr.5: 69381988-69381988 on Build GRCh38
rs369055005 Chr.5: 69381905-69381905 on Build GRCh38
rs146098532 Chr.5: 69381966-69381966 on Build GRCh38
rs747635308 Chr.5: 69382009-69382009 on Build GRCh38
rs756921229 Chr.5: 69381950-69381950 on Build GRCh38
rs768589147 Chr.5: 69381996-69381996 on Build GRCh38
rs777138649 Chr.5: 69382034-69382034 on Build GRCh38
rs142829528 Chr.5: 69381949-69381949 on Build GRCh38
rs373675696 Chr.5: 69381990-69381990 on Build GRCh38
rs748245370 Chr.5: 69381951-69381951 on Build GRCh38
rs745641746 Chr.5: 69381974-69381974 on Build GRCh38
rs199971023 Chr.5: 69381906-69381906 on Build GRCh38
rs773810590 Chr.5: 69381997-69381997 on Build GRCh38
rs771734393 Chr.5: 69382012-69382012 on Build GRCh38
rs773578866 Chr.5: 69381915-69381915 on Build GRCh38
rs779762811 Chr.5: 69382077-69382077 on Build GRCh38
rs772781432 Chr.5: 69382025-69382025 on Build GRCh38
rs755555015 Chr.5: 69381936-69381936 on Build GRCh38
rs753665575 Chr.5: 69382079-69382079 on Build GRCh38
rs1051955562 Chr.5: 69381918-69381918 on Build GRCh38
rs754452974 Chr.5: 69381923-69381923 on Build GRCh38
rs765858019 Chr.5: 69382056-69382056 on Build GRCh38
rs369510585 Chr.5: 69381922-69381922 on Build GRCh38
rs184637090 Chr.5: 69381941-69381941 on Build GRCh38
rs766790790 Chr.5: 69382033-69382033 on Build GRCh38
rs374791081 Chr.5: 69381964-69381964 on Build GRCh38

View Full Product Details