Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33937247-33937506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXFP3
Entrez Gene ID
51289
Gene Name
relaxin/insulin like family peptide receptor 3
Gene Aliases
GPCR135, RLN3R1, RXFPR3, SALPR
UniGene
Hs.170146
Gene Chromosome Location
Chr.5: 33936386-33938918 on Build GRCh38
Transcripts
NM_016568.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738045 c.639G>T p.S213S Chr.5: 33937379-33937379 on Build GRCh38
3947447 c.756G>T p.P252P Chr.5: 33937496-33937496 on Build GRCh38
1264704 c.676G>A p.A226T Chr.5: 33937416-33937416 on Build GRCh38
6103864 c.592C>A p.R198R Chr.5: 33937332-33937332 on Build GRCh38
3697214 c.676G>T p.A226S Chr.5: 33937416-33937416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759357991 Chr.5: 33937276-33937277 on Build GRCh38
rs762511560 Chr.5: 33937301-33937301 on Build GRCh38
rs767232942 Chr.5: 33937345-33937345 on Build GRCh38
rs758425756 Chr.5: 33937423-33937423 on Build GRCh38
rs770813828 Chr.5: 33937267-33937268 on Build GRCh38
rs776074089 Chr.5: 33937312-33937312 on Build GRCh38
rs145602095 Chr.5: 33937488-33937488 on Build GRCh38
rs62350867 Chr.5: 33937309-33937309 on Build GRCh38
rs754956127 Chr.5: 33937433-33937433 on Build GRCh38
rs763280924 Chr.5: 33937329-33937329 on Build GRCh38
rs1050898469 Chr.5: 33937279-33937279 on Build GRCh38
rs762227783 Chr.5: 33937414-33937414 on Build GRCh38
rs374524110 Chr.5: 33937367-33937367 on Build GRCh38
rs200499016 Chr.5: 33937453-33937453 on Build GRCh38
rs529072137 Chr.5: 33937269-33937269 on Build GRCh38
rs547132118 Chr.5: 33937454-33937454 on Build GRCh38
rs747951202 Chr.5: 33937362-33937362 on Build GRCh38
rs267600603 Chr.5: 33937290-33937290 on Build GRCh38
rs746895228 Chr.5: 33937339-33937339 on Build GRCh38
rs1035788535 Chr.5: 33937467-33937467 on Build GRCh38
rs760814848 Chr.5: 33937328-33937345 on Build GRCh38
rs1023157563 Chr.5: 33937415-33937415 on Build GRCh38
rs200520186 Chr.5: 33937379-33937379 on Build GRCh38
rs771024289 Chr.5: 33937477-33937477 on Build GRCh38
rs200828663 Chr.5: 33937326-33937326 on Build GRCh38
rs905914100 Chr.5: 33937441-33937441 on Build GRCh38
rs562455881 Chr.5: 33937298-33937298 on Build GRCh38
rs754795557 Chr.5: 33937350-33937350 on Build GRCh38
rs531626591 Chr.5: 33937300-33937300 on Build GRCh38
rs781004172 Chr.5: 33937355-33937355 on Build GRCh38
rs199540993 Chr.5: 33937461-33937461 on Build GRCh38
rs748753459 Chr.5: 33937369-33937369 on Build GRCh38
rs754146433 Chr.5: 33937315-33937315 on Build GRCh38
rs768204709 Chr.5: 33937307-33937307 on Build GRCh38
rs768378041 Chr.5: 33937284-33937284 on Build GRCh38
rs147626527 Chr.5: 33937388-33937388 on Build GRCh38
rs757464013 Chr.5: 33937320-33937320 on Build GRCh38
rs752361735 Chr.5: 33937395-33937396 on Build GRCh38
rs779975881 Chr.5: 33937333-33937333 on Build GRCh38
rs758504033 Chr.5: 33937328-33937328 on Build GRCh38
rs965528796 Chr.5: 33937351-33937351 on Build GRCh38
rs776285970 Chr.5: 33937285-33937285 on Build GRCh38
rs201917378 Chr.5: 33937314-33937314 on Build GRCh38
rs367807433 Chr.5: 33937427-33937427 on Build GRCh38
rs776911305 Chr.5: 33937400-33937400 on Build GRCh38
rs750530756 Chr.5: 33937327-33937327 on Build GRCh38
rs765447933 Chr.5: 33937418-33937418 on Build GRCh38
rs757382584 Chr.5: 33937268-33937268 on Build GRCh38
rs776164315 Chr.5: 33937380-33937380 on Build GRCh38
rs770508250 Chr.5: 33937375-33937375 on Build GRCh38
rs548724371 Chr.5: 33937277-33937277 on Build GRCh38
rs761225440 Chr.5: 33937387-33937387 on Build GRCh38
rs750597170 Chr.5: 33937422-33937422 on Build GRCh38
rs145194382 Chr.5: 33937297-33937297 on Build GRCh38
rs536064158 Chr.5: 33937481-33937481 on Build GRCh38
rs769389469 Chr.5: 33937364-33937364 on Build GRCh38
rs774249278 Chr.5: 33937272-33937273 on Build GRCh38
rs770598320 Chr.5: 33937468-33937468 on Build GRCh38
rs781092269 Chr.5: 33937434-33937434 on Build GRCh38
rs984011075 Chr.5: 33937431-33937431 on Build GRCh38
rs745931932 Chr.5: 33937271-33937271 on Build GRCh38
rs751604901 Chr.5: 33937429-33937429 on Build GRCh38
rs747960415 Chr.5: 33937451-33937451 on Build GRCh38

View Full Product Details