Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89778034-89778305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MESP2
Entrez Gene ID
145873
Gene Name
mesoderm posterior bHLH transcription factor 2
Gene Aliases
SCDO2, bHLHc6
UniGene
Hs.37311
Gene Chromosome Location
Chr.15: 89776358-89778754 on Build GRCh38
Transcripts
NM_001039958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647864 c.931T>G p.S311A Chr.15: 89778071-89778071 on Build GRCh38
5700725 c.1080C>T p.A360A Chr.15: 89778220-89778220 on Build GRCh38
5850049 c.941C>G p.P314R Chr.15: 89778081-89778081 on Build GRCh38
3765837 c.1086C>G p.F362L Chr.15: 89778226-89778226 on Build GRCh38
5882640 c.947C>G p.P316R Chr.15: 89778087-89778087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763376959 Chr.15: 89778214-89778214 on Build GRCh38
rs201495246 Chr.15: 89778196-89778196 on Build GRCh38
rs767430602 Chr.15: 89778221-89778221 on Build GRCh38
rs879095532 Chr.15: 89778154-89778154 on Build GRCh38
rs762014488 Chr.15: 89778220-89778220 on Build GRCh38
rs564292487 Chr.15: 89778213-89778213 on Build GRCh38
rs917606111 Chr.15: 89778082-89778082 on Build GRCh38
rs546107097 Chr.15: 89778212-89778212 on Build GRCh38
rs923308382 Chr.15: 89778278-89778279 on Build GRCh38
rs777196250 Chr.15: 89778210-89778210 on Build GRCh38
rs199514997 Chr.15: 89778184-89778184 on Build GRCh38
rs779823676 Chr.15: 89778081-89778081 on Build GRCh38
rs760177391 Chr.15: 89778120-89778120 on Build GRCh38
rs376942190 Chr.15: 89778142-89778142 on Build GRCh38
rs765449941 Chr.15: 89778144-89778144 on Build GRCh38
rs370227491 Chr.15: 89778157-89778157 on Build GRCh38
rs746574351 Chr.15: 89778199-89778199 on Build GRCh38
rs752972579 Chr.15: 89778145-89778145 on Build GRCh38
rs751673727 Chr.15: 89778072-89778072 on Build GRCh38
rs775941947 Chr.15: 89778199-89778200 on Build GRCh38
rs754807279 Chr.15: 89778176-89778176 on Build GRCh38
rs758115773 Chr.15: 89778190-89778190 on Build GRCh38
rs771471316 Chr.15: 89778125-89778125 on Build GRCh38
rs777939697 Chr.15: 89778065-89778065 on Build GRCh38
rs772586292 Chr.15: 89778153-89778153 on Build GRCh38
rs771586079 Chr.15: 89778115-89778115 on Build GRCh38
rs374475842 Chr.15: 89778167-89778167 on Build GRCh38
rs71647810 Chr.15: 89778192-89778192 on Build GRCh38
rs768285654 Chr.15: 89778106-89778106 on Build GRCh38
rs776986584 Chr.15: 89778127-89778127 on Build GRCh38
rs774665542 Chr.15: 89778218-89778218 on Build GRCh38
rs765218369 Chr.15: 89778262-89778262 on Build GRCh38
rs897440052 Chr.15: 89778160-89778160 on Build GRCh38
rs747926651 Chr.15: 89778187-89778187 on Build GRCh38
rs753827969 Chr.15: 89778229-89778229 on Build GRCh38
rs905495693 Chr.15: 89778104-89778104 on Build GRCh38
rs778659855 Chr.15: 89778107-89778107 on Build GRCh38
rs925442555 Chr.15: 89778069-89778069 on Build GRCh38
rs770301033 Chr.15: 89778203-89778203 on Build GRCh38
rs763321859 Chr.15: 89778150-89778150 on Build GRCh38
rs756185244 Chr.15: 89778080-89778080 on Build GRCh38
rs757634059 Chr.15: 89778088-89778089 on Build GRCh38
rs372981270 Chr.15: 89778116-89778116 on Build GRCh38
rs376465704 Chr.15: 89778217-89778217 on Build GRCh38
rs780062393 Chr.15: 89778158-89778158 on Build GRCh38
rs779274067 Chr.15: 89778142-89778143 on Build GRCh38
rs764403618 Chr.15: 89778151-89778151 on Build GRCh38
rs752531314 Chr.15: 89778266-89778266 on Build GRCh38
rs754930000 Chr.15: 89778244-89778244 on Build GRCh38
rs752665246 Chr.15: 89778097-89778097 on Build GRCh38
rs777428484 Chr.15: 89778280-89778280 on Build GRCh38
rs758170634 Chr.15: 89778267-89778267 on Build GRCh38
rs746380436 Chr.15: 89778145-89778146 on Build GRCh38
rs747732638 Chr.15: 89778113-89778113 on Build GRCh38
rs1044843208 Chr.15: 89778118-89778118 on Build GRCh38

View Full Product Details