Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49999367-49999608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM6
Entrez Gene ID
10180
Gene Name
RNA binding motif protein 6
Gene Aliases
3G2, DEF-3, DEF3, HLC-11, NY-LU-12, g16
UniGene
Hs.696735
Gene Chromosome Location
Chr.3: 49940038-50077252 on Build GRCh38
Transcripts
NM_001167582.1 NM_005777.2 XM_005264784.1 XM_005264785.1 XM_005264786.1 XM_005264787.2 XM_006712916.1 XM_017005496.1 XM_017005497.1 XM_017005498.1 XM_017005499.1 XM_017005500.1 XM_017005501.1 XM_017005502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7341648 c.1555C>T p.Q519* Chr.3: 49999511-49999511 on Build GRCh38
7090153 c.1521C>A p.L507L Chr.3: 49999477-49999477 on Build GRCh38
6764191 c.1537G>A p.G513R Chr.3: 49999493-49999493 on Build GRCh38
2850943 c.1512G>A p.E504E Chr.3: 49999468-49999468 on Build GRCh38
6252461 c.1507G>T p.V503L Chr.3: 49999463-49999463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9851274 Chr.3: 49999399-49999399 on Build GRCh38
rs376522104 Chr.3: 49999456-49999456 on Build GRCh38
rs760316093 Chr.3: 49999395-49999395 on Build GRCh38
rs761508462 Chr.3: 49999414-49999414 on Build GRCh38
rs778986011 Chr.3: 49999505-49999505 on Build GRCh38
rs756504858 Chr.3: 49999468-49999468 on Build GRCh38
rs371026751 Chr.3: 49999561-49999561 on Build GRCh38
rs200764628 Chr.3: 49999529-49999529 on Build GRCh38
rs755094107 Chr.3: 49999457-49999457 on Build GRCh38
rs768381629 Chr.3: 49999553-49999553 on Build GRCh38
rs778187801 Chr.3: 49999471-49999471 on Build GRCh38
rs776458536 Chr.3: 49999554-49999554 on Build GRCh38
rs201026641 Chr.3: 49999431-49999431 on Build GRCh38
rs749511045 Chr.3: 49999490-49999490 on Build GRCh38
rs755567450 Chr.3: 49999434-49999434 on Build GRCh38
rs763943036 Chr.3: 49999398-49999398 on Build GRCh38
rs947675408 Chr.3: 49999477-49999477 on Build GRCh38
rs781362481 Chr.3: 49999555-49999555 on Build GRCh38
rs988372765 Chr.3: 49999475-49999475 on Build GRCh38
rs768080175 Chr.3: 49999435-49999435 on Build GRCh38
rs746949622 Chr.3: 49999551-49999551 on Build GRCh38
rs764883531 Chr.3: 49999415-49999415 on Build GRCh38
rs369920008 Chr.3: 49999500-49999500 on Build GRCh38
rs766190321 Chr.3: 49999553-49999553 on Build GRCh38
rs751521089 Chr.3: 49999553-49999554 on Build GRCh38
rs373771738 Chr.3: 49999528-49999528 on Build GRCh38
rs745889848 Chr.3: 49999506-49999506 on Build GRCh38
rs772865652 Chr.3: 49999563-49999563 on Build GRCh38
rs776202680 Chr.3: 49999405-49999405 on Build GRCh38

View Full Product Details