Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56122773-56123032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8H3
Entrez Gene ID
390152
Gene Name
olfactory receptor family 8 subfamily H member 3
Gene Aliases
OR11-172
UniGene
Hs.553745
Gene Chromosome Location
Chr.11: 56122373-56123311 on Build GRCh38
Transcripts
NM_001005201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365627 c.622A>G p.M208V Chr.11: 56122994-56122994 on Build GRCh38
330821 c.423C>A p.C141* Chr.11: 56122795-56122795 on Build GRCh38
75791 c.520A>T p.I174F Chr.11: 56122892-56122892 on Build GRCh38
346986 c.657G>T p.V219V Chr.11: 56123029-56123029 on Build GRCh38
4407927 c.420C>G p.L140L Chr.11: 56122792-56122792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377459055 Chr.11: 56122979-56122979 on Build GRCh38
rs887860542 Chr.11: 56122847-56122847 on Build GRCh38
rs17531522 Chr.11: 56122973-56122973 on Build GRCh38
rs763705738 Chr.11: 56122994-56122994 on Build GRCh38
rs769092958 Chr.11: 56122949-56122949 on Build GRCh38
rs140813125 Chr.11: 56122837-56122837 on Build GRCh38
rs61743843 Chr.11: 56122821-56122821 on Build GRCh38
rs148145935 Chr.11: 56122978-56122978 on Build GRCh38
rs754898397 Chr.11: 56122889-56122889 on Build GRCh38
rs371099411 Chr.11: 56122877-56122877 on Build GRCh38
rs1015327404 Chr.11: 56122878-56122878 on Build GRCh38
rs61743838 Chr.11: 56122844-56122844 on Build GRCh38
rs893615303 Chr.11: 56122835-56122835 on Build GRCh38
rs868650876 Chr.11: 56122993-56122993 on Build GRCh38
rs748295322 Chr.11: 56122960-56122960 on Build GRCh38
rs201126799 Chr.11: 56122827-56122827 on Build GRCh38
rs552568669 Chr.11: 56122815-56122815 on Build GRCh38
rs780318369 Chr.11: 56122934-56122934 on Build GRCh38
rs546367458 Chr.11: 56122914-56122914 on Build GRCh38
rs752301022 Chr.11: 56122909-56122909 on Build GRCh38
rs755850706 Chr.11: 56122802-56122802 on Build GRCh38
rs141920018 Chr.11: 56122980-56122980 on Build GRCh38
rs375397632 Chr.11: 56122950-56122950 on Build GRCh38
rs754452341 Chr.11: 56122798-56122798 on Build GRCh38
rs748964769 Chr.11: 56122917-56122917 on Build GRCh38
rs541593719 Chr.11: 56122849-56122849 on Build GRCh38
rs769988999 Chr.11: 56122972-56122972 on Build GRCh38
rs756258601 Chr.11: 56122903-56122903 on Build GRCh38
rs573660406 Chr.11: 56122843-56122843 on Build GRCh38
rs145288913 Chr.11: 56122895-56122895 on Build GRCh38
rs772448463 Chr.11: 56122833-56122833 on Build GRCh38
rs747501578 Chr.11: 56122945-56122945 on Build GRCh38
rs140934188 Chr.11: 56122963-56122963 on Build GRCh38
rs758726988 Chr.11: 56122918-56122918 on Build GRCh38
rs780445547 Chr.11: 56123006-56123006 on Build GRCh38
rs61746318 Chr.11: 56122880-56122880 on Build GRCh38
rs61743852 Chr.11: 56122797-56122797 on Build GRCh38
rs1044965007 Chr.11: 56122863-56122863 on Build GRCh38
rs776011698 Chr.11: 56122834-56122834 on Build GRCh38
rs924244076 Chr.11: 56122805-56122805 on Build GRCh38
rs762861968 Chr.11: 56122860-56122860 on Build GRCh38
rs753493595 Chr.11: 56123000-56123000 on Build GRCh38
rs540879053 Chr.11: 56122799-56122799 on Build GRCh38
rs760208900 Chr.11: 56122991-56122991 on Build GRCh38
rs756882449 Chr.11: 56123001-56123001 on Build GRCh38
rs767601042 Chr.11: 56122982-56122982 on Build GRCh38
rs772985957 Chr.11: 56122852-56122852 on Build GRCh38
rs752243837 Chr.11: 56122989-56122989 on Build GRCh38
rs768742706 Chr.11: 56122842-56122842 on Build GRCh38
rs755794918 Chr.11: 56122912-56122912 on Build GRCh38
rs78856925 Chr.11: 56122879-56122879 on Build GRCh38
rs754823017 Chr.11: 56122923-56122924 on Build GRCh38
rs146422829 Chr.11: 56122956-56122956 on Build GRCh38

View Full Product Details