Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37395953-37396226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRC
Entrez Gene ID
6714
Gene Name
SRC proto-oncogene, non-receptor tyrosine kinase
Gene Aliases
ASV, SRC1, THC6, c-SRC, p60-Src
UniGene
Hs.195659
Gene Chromosome Location
Chr.20: 37344685-37405432 on Build GRCh38
Transcripts
NM_005417.4 NM_198291.2 XM_011529013.2 XM_017028024.1 XM_017028025.1 XM_017028026.1 XM_017028027.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6973721 c.556G>A p.A186T Chr.20: 37396164-37396164 on Build GRCh38
6938320 c.594G>T p.K198N Chr.20: 37396202-37396202 on Build GRCh38
6370054 c.617A>G p.K206R Chr.20: 37396225-37396225 on Build GRCh38
7297286 c.588C>A p.N196K Chr.20: 37396196-37396196 on Build GRCh38
6368595 c.568T>C p.S190P Chr.20: 37396176-37396176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045310538 Chr.20: 37396094-37396094 on Build GRCh38
rs745663583 Chr.20: 37396148-37396148 on Build GRCh38
rs201170704 Chr.20: 37396169-37396169 on Build GRCh38
rs556995998 Chr.20: 37396052-37396052 on Build GRCh38
rs201680915 Chr.20: 37396134-37396134 on Build GRCh38
rs1032372078 Chr.20: 37396144-37396144 on Build GRCh38
rs1006244613 Chr.20: 37396062-37396062 on Build GRCh38
rs6012117 Chr.20: 37396157-37396157 on Build GRCh38
rs750781383 Chr.20: 37396126-37396126 on Build GRCh38
rs931476968 Chr.20: 37396042-37396042 on Build GRCh38
rs781751002 Chr.20: 37396198-37396198 on Build GRCh38
rs753404366 Chr.20: 37396204-37396204 on Build GRCh38
rs535137779 Chr.20: 37396017-37396017 on Build GRCh38
rs922353212 Chr.20: 37396026-37396027 on Build GRCh38
rs929682222 Chr.20: 37396039-37396039 on Build GRCh38
rs765391344 Chr.20: 37396125-37396125 on Build GRCh38
rs1035790642 Chr.20: 37396108-37396108 on Build GRCh38
rs770739484 Chr.20: 37396143-37396143 on Build GRCh38
rs748762036 Chr.20: 37396141-37396141 on Build GRCh38
rs747622121 Chr.20: 37396137-37396137 on Build GRCh38
rs1047694063 Chr.20: 37396051-37396051 on Build GRCh38
rs11474889 Chr.20: 37395985-37395985 on Build GRCh38
rs905648791 Chr.20: 37396071-37396071 on Build GRCh38
rs755567292 Chr.20: 37396197-37396197 on Build GRCh38
rs774600805 Chr.20: 37396115-37396116 on Build GRCh38
rs912801439 Chr.20: 37395984-37395984 on Build GRCh38
rs777477625 Chr.20: 37396140-37396140 on Build GRCh38
rs1040693329 Chr.20: 37396014-37396014 on Build GRCh38
rs180818054 Chr.20: 37396114-37396114 on Build GRCh38
rs760418101 Chr.20: 37396116-37396116 on Build GRCh38
rs75363208 Chr.20: 37396168-37396168 on Build GRCh38
rs763085496 Chr.20: 37396181-37396181 on Build GRCh38
rs367842964 Chr.20: 37396115-37396115 on Build GRCh38
rs776712117 Chr.20: 37396158-37396158 on Build GRCh38
rs766227683 Chr.20: 37396184-37396189 on Build GRCh38
rs757522887 Chr.20: 37396122-37396122 on Build GRCh38
rs541715085 Chr.20: 37396145-37396145 on Build GRCh38
rs140526991 Chr.20: 37396008-37396008 on Build GRCh38
rs575291811 Chr.20: 37396074-37396074 on Build GRCh38
rs761837421 Chr.20: 37396166-37396166 on Build GRCh38
rs397897665 Chr.20: 37395987-37395987 on Build GRCh38
rs750597214 Chr.20: 37396178-37396178 on Build GRCh38
rs200185588 Chr.20: 37396150-37396150 on Build GRCh38
rs1037304707 Chr.20: 37396012-37396012 on Build GRCh38
rs559832663 Chr.20: 37396149-37396149 on Build GRCh38
rs549125627 Chr.20: 37396096-37396096 on Build GRCh38
rs897276448 Chr.20: 37396040-37396040 on Build GRCh38
rs372806161 Chr.20: 37396133-37396133 on Build GRCh38
rs766810850 Chr.20: 37396190-37396190 on Build GRCh38
rs368943243 Chr.20: 37396116-37396116 on Build GRCh38
rs764225167 Chr.20: 37396118-37396118 on Build GRCh38
rs140451827 Chr.20: 37396196-37396196 on Build GRCh38
rs370767993 Chr.20: 37396139-37396139 on Build GRCh38

View Full Product Details