Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105881262-105881535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCK2
Entrez Gene ID
8440
Gene Name
NCK adaptor protein 2
Gene Aliases
GRB4, NCKbeta
UniGene
Hs.529244
Gene Chromosome Location
Chr.2: 105744649-105894274 on Build GRCh38
Transcripts
NM_001004720.2 NM_001004722.3 NM_003581.4 XM_006712797.3 XM_011511991.2 XM_011511992.2 XM_017005103.1 XM_017005104.1 XM_017005105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3564679 c.392C>T p.S131F Chr.2: 105881493-105881493 on Build GRCh38
1564879 c.256G>A p.A86T Chr.2: 105881357-105881357 on Build GRCh38
6152928 c.313G>A p.G105S Chr.2: 105881414-105881414 on Build GRCh38
4598049 c.331G>A p.D111N Chr.2: 105881432-105881432 on Build GRCh38
6722935 c.431G>A p.C144Y Chr.2: 105881532-105881532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762019822 Chr.2: 105881467-105881467 on Build GRCh38
rs921802614 Chr.2: 105881300-105881300 on Build GRCh38
rs747636275 Chr.2: 105881443-105881443 on Build GRCh38
rs140384294 Chr.2: 105881298-105881298 on Build GRCh38
rs973651345 Chr.2: 105881369-105881369 on Build GRCh38
rs776872198 Chr.2: 105881385-105881385 on Build GRCh38
rs748935902 Chr.2: 105881361-105881361 on Build GRCh38
rs761674486 Chr.2: 105881408-105881408 on Build GRCh38
rs778588738 Chr.2: 105881439-105881439 on Build GRCh38
rs147879012 Chr.2: 105881474-105881474 on Build GRCh38
rs753586078 Chr.2: 105881433-105881433 on Build GRCh38
rs764141400 Chr.2: 105881320-105881320 on Build GRCh38
rs781754645 Chr.2: 105881444-105881444 on Build GRCh38
rs757980179 Chr.2: 105881495-105881495 on Build GRCh38
rs138508918 Chr.2: 105881362-105881362 on Build GRCh38
rs919479658 Chr.2: 105881431-105881431 on Build GRCh38
rs780761768 Chr.2: 105881284-105881284 on Build GRCh38
rs569250884 Chr.2: 105881448-105881448 on Build GRCh38
rs766256305 Chr.2: 105881432-105881432 on Build GRCh38
rs755994999 Chr.2: 105881418-105881418 on Build GRCh38
rs765438756 Chr.2: 105881297-105881297 on Build GRCh38
rs757182579 Chr.2: 105881333-105881333 on Build GRCh38
rs764192536 Chr.2: 105881402-105881402 on Build GRCh38
rs375422663 Chr.2: 105881478-105881478 on Build GRCh38
rs747525415 Chr.2: 105881294-105881295 on Build GRCh38
rs770165493 Chr.2: 105881446-105881446 on Build GRCh38
rs11553854 Chr.2: 105881509-105881509 on Build GRCh38
rs763082202 Chr.2: 105881310-105881310 on Build GRCh38
rs770047813 Chr.2: 105881387-105881387 on Build GRCh38
rs117970405 Chr.2: 105881374-105881374 on Build GRCh38
rs772032136 Chr.2: 105881468-105881468 on Build GRCh38
rs1002685698 Chr.2: 105881285-105881285 on Build GRCh38
rs113420409 Chr.2: 105881473-105881473 on Build GRCh38
rs752283243 Chr.2: 105881488-105881488 on Build GRCh38
rs777123504 Chr.2: 105881511-105881511 on Build GRCh38
rs201406196 Chr.2: 105881388-105881388 on Build GRCh38
rs142427776 Chr.2: 105881359-105881359 on Build GRCh38
rs763135850 Chr.2: 105881397-105881397 on Build GRCh38
rs755727286 Chr.2: 105881354-105881354 on Build GRCh38
rs150904549 Chr.2: 105881389-105881389 on Build GRCh38
rs747500810 Chr.2: 105881376-105881376 on Build GRCh38
rs913136866 Chr.2: 105881358-105881358 on Build GRCh38
rs937139915 Chr.2: 105881508-105881508 on Build GRCh38
rs774367973 Chr.2: 105881406-105881406 on Build GRCh38
rs754635258 Chr.2: 105881434-105881434 on Build GRCh38
rs771417618 Chr.2: 105881379-105881379 on Build GRCh38
rs751541163 Chr.2: 105881330-105881330 on Build GRCh38
rs150031257 Chr.2: 105881416-105881416 on Build GRCh38
rs749536605 Chr.2: 105881462-105881462 on Build GRCh38
rs767240597 Chr.2: 105881340-105881340 on Build GRCh38
rs753636963 Chr.2: 105881477-105881477 on Build GRCh38
rs199727155 Chr.2: 105881461-105881461 on Build GRCh38
rs750177299 Chr.2: 105881341-105881341 on Build GRCh38
rs746174788 Chr.2: 105881386-105881386 on Build GRCh38
rs767425900 Chr.2: 105881413-105881413 on Build GRCh38
rs774422971 Chr.2: 105881464-105881464 on Build GRCh38
rs200636557 Chr.2: 105881481-105881481 on Build GRCh38

View Full Product Details