Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37495819-37496058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1E
Entrez Gene ID
64425
Gene Name
RNA polymerase I subunit E
Gene Aliases
PAF53, PRAF1
UniGene
Hs.591087
Gene Chromosome Location
Chr.9: 37485935-37503697 on Build GRCh38
Transcripts
NM_001282766.1 NM_022490.2 XM_005251547.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6752308 c.902C>T p.T301M Chr.9: 37495950-37495950 on Build GRCh38
487461 c.711C>T p.N237N Chr.9: 37495945-37495945 on Build GRCh38
5977245 c.522G>T p.L174L Chr.9: 37495966-37495966 on Build GRCh38
7174717 c.692C>T p.P231L Chr.9: 37495926-37495926 on Build GRCh38
7402236 c.514G>T p.E172* Chr.9: 37495958-37495958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150425999 Chr.9: 37495950-37495950 on Build GRCh38
rs774754328 Chr.9: 37495901-37495901 on Build GRCh38
rs772507938 Chr.9: 37496009-37496009 on Build GRCh38
rs986781803 Chr.9: 37495960-37495960 on Build GRCh38
rs762322864 Chr.9: 37496025-37496025 on Build GRCh38
rs750335393 Chr.9: 37495990-37495990 on Build GRCh38
rs746246750 Chr.9: 37496005-37496005 on Build GRCh38
rs773068402 Chr.9: 37495945-37495945 on Build GRCh38
rs372254576 Chr.9: 37496018-37496018 on Build GRCh38
rs756249765 Chr.9: 37495984-37495984 on Build GRCh38
rs367830004 Chr.9: 37496016-37496016 on Build GRCh38
rs764826753 Chr.9: 37495944-37495944 on Build GRCh38
rs771236932 Chr.9: 37495897-37495897 on Build GRCh38
rs115817892 Chr.9: 37495948-37495948 on Build GRCh38
rs202057503 Chr.9: 37495986-37495986 on Build GRCh38
rs16934203 Chr.9: 37495902-37495902 on Build GRCh38
rs748549089 Chr.9: 37495989-37495989 on Build GRCh38
rs761422375 Chr.9: 37495942-37495942 on Build GRCh38
rs767451399 Chr.9: 37495951-37495951 on Build GRCh38
rs772665721 Chr.9: 37495908-37495908 on Build GRCh38
rs752614900 Chr.9: 37495977-37495977 on Build GRCh38
rs190430559 Chr.9: 37495990-37495990 on Build GRCh38
rs773811632 Chr.9: 37496033-37496033 on Build GRCh38
rs749505546 Chr.9: 37495892-37495892 on Build GRCh38
rs756125217 Chr.9: 37495855-37495855 on Build GRCh38
rs776072238 Chr.9: 37495921-37495921 on Build GRCh38
rs777797212 Chr.9: 37495985-37495985 on Build GRCh38
rs772828883 Chr.9: 37496022-37496022 on Build GRCh38
rs554470455 Chr.9: 37495862-37495862 on Build GRCh38
rs780763830 Chr.9: 37496012-37496012 on Build GRCh38
rs545783299 Chr.9: 37496001-37496001 on Build GRCh38
rs376803930 Chr.9: 37496026-37496026 on Build GRCh38

View Full Product Details