Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49297267-49297524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927267
Entrez Gene ID
101927267
Gene Name
uncharacterized LOC101927267
Gene Aliases
-
UniGene
Hs.640101
Gene Chromosome Location
Chr.12: 49292631-49324576 on Build GRCh38
Transcripts
NR_120449.1 NM_006262.3 XM_005269025.1

Gene Symbol
PRPH
Entrez Gene ID
5630
Gene Name
peripherin
Gene Aliases
NEF4, PRPH1
UniGene
Hs.37044
Gene Chromosome Location
Chr.12: 49295013-49298698 on Build GRCh38
Transcripts
NR_120449.1 NM_006262.3 XM_005269025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1171909 c.1048G>T p.E350* Chr.12: 49297408-49297408 on Build GRCh38
693620 c.1160T>C p.M387T Chr.12: 49297520-49297520 on Build GRCh38
3931642 c.1071C>G p.G357G Chr.12: 49297431-49297431 on Build GRCh38
2007790 c.1046T>C p.L349P Chr.12: 49297406-49297406 on Build GRCh38
3792648 c.1097G>C p.R366P Chr.12: 49297457-49297457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571876859 Chr.12: 49297371-49297371 on Build GRCh38
rs796364482 Chr.12: 49297442-49297442 on Build GRCh38
rs549558070 Chr.12: 49297306-49297306 on Build GRCh38
rs774487913 Chr.12: 49297431-49297431 on Build GRCh38
rs777418543 Chr.12: 49297418-49297418 on Build GRCh38
rs753255815 Chr.12: 49297393-49297393 on Build GRCh38
rs747034268 Chr.12: 49297419-49297419 on Build GRCh38
rs748522272 Chr.12: 49297433-49297433 on Build GRCh38
rs754725165 Chr.12: 49297398-49297398 on Build GRCh38
rs768846672 Chr.12: 49297359-49297359 on Build GRCh38
rs751294625 Chr.12: 49297456-49297456 on Build GRCh38
rs762359109 Chr.12: 49297303-49297303 on Build GRCh38
rs764513374 Chr.12: 49297292-49297292 on Build GRCh38
rs73112143 Chr.12: 49297467-49297467 on Build GRCh38
rs765957049 Chr.12: 49297311-49297311 on Build GRCh38
rs989014427 Chr.12: 49297430-49297430 on Build GRCh38
rs764932638 Chr.12: 49297412-49297412 on Build GRCh38
rs766773435 Chr.12: 49297440-49297440 on Build GRCh38
rs752456220 Chr.12: 49297449-49297449 on Build GRCh38
rs761980917 Chr.12: 49297380-49297381 on Build GRCh38
rs753580540 Chr.12: 49297324-49297324 on Build GRCh38
rs773707575 Chr.12: 49297369-49297369 on Build GRCh38
rs758211963 Chr.12: 49297415-49297415 on Build GRCh38
rs775431866 Chr.12: 49297348-49297348 on Build GRCh38
rs1012240852 Chr.12: 49297428-49297428 on Build GRCh38
rs762690778 Chr.12: 49297450-49297450 on Build GRCh38
rs537023141 Chr.12: 49297413-49297413 on Build GRCh38
rs535948895 Chr.12: 49297474-49297474 on Build GRCh38
rs749320316 Chr.12: 49297427-49297427 on Build GRCh38
rs768899850 Chr.12: 49297429-49297429 on Build GRCh38
rs867888351 Chr.12: 49297329-49297329 on Build GRCh38
rs62636517 Chr.12: 49297443-49297443 on Build GRCh38
rs367981648 Chr.12: 49297464-49297464 on Build GRCh38
rs751894948 Chr.12: 49297301-49297301 on Build GRCh38
rs59827092 Chr.12: 49297468-49297468 on Build GRCh38
rs757243807 Chr.12: 49297457-49297457 on Build GRCh38
rs1014087794 Chr.12: 49297299-49297299 on Build GRCh38
rs778891245 Chr.12: 49297334-49297334 on Build GRCh38
rs774723000 Chr.12: 49297360-49297360 on Build GRCh38
rs372792711 Chr.12: 49297475-49297475 on Build GRCh38
rs528720936 Chr.12: 49297337-49297337 on Build GRCh38
rs748432214 Chr.12: 49297503-49297503 on Build GRCh38
rs370910135 Chr.12: 49297439-49297439 on Build GRCh38
rs373684818 Chr.12: 49297342-49297342 on Build GRCh38
rs773441319 Chr.12: 49297435-49297435 on Build GRCh38
rs552035032 Chr.12: 49297351-49297351 on Build GRCh38
rs980280041 Chr.12: 49297363-49297363 on Build GRCh38
rs765336123 Chr.12: 49297413-49297413 on Build GRCh38
rs764997995 Chr.12: 49297445-49297445 on Build GRCh38
rs763994010 Chr.12: 49297453-49297453 on Build GRCh38
rs768013983 Chr.12: 49297364-49297364 on Build GRCh38
rs757115552 Chr.12: 49297423-49297423 on Build GRCh38
rs765980006 Chr.12: 49297374-49297374 on Build GRCh38
rs369481202 Chr.12: 49297335-49297335 on Build GRCh38
rs759075246 Chr.12: 49297444-49297444 on Build GRCh38
rs778946909 Chr.12: 49297497-49297497 on Build GRCh38
rs772660388 Chr.12: 49297380-49297389 on Build GRCh38
rs200519578 Chr.12: 49297310-49297310 on Build GRCh38
rs376923557 Chr.12: 49297330-49297330 on Build GRCh38
rs377665045 Chr.12: 49297406-49297406 on Build GRCh38
rs375933306 Chr.12: 49297347-49297347 on Build GRCh38
rs1038199227 Chr.12: 49297308-49297308 on Build GRCh38

View Full Product Details