Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 176043489-176043762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFWD2
Entrez Gene ID
64326
Gene Name
ring finger and WD repeat domain 2
Gene Aliases
COP1, RNF200
UniGene
Hs.523744
Gene Chromosome Location
Chr.1: 175944826-176207279 on Build GRCh38
Transcripts
NM_001001740.3 NM_001286644.1 NM_022457.6 XM_005245447.2 XM_005245448.2 XM_006711487.2 XM_011509881.1 XM_017002059.1 XM_017002060.1 XM_017002061.1 XM_017002062.1 XM_017002063.1 XM_017002064.1 XM_017002065.1 XM_017002066.1 XM_017002067.1 XM_017002068.1 XM_017002069.1 XM_017002070.1 XM_017002071.1 XM_017002072.1 XM_017002073.1 XM_017002074.1 XM_017002075.1 XM_017002076.1 XM_017002077.1 XM_017002078.1 XM_017002079.1 XM_017002080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849332 c.1530+5A>C p.? Chr.1: 176043705-176043705 on Build GRCh38
5787411 c.1508G>T p.G503V Chr.1: 176043732-176043732 on Build GRCh38
6921485 c.1521G>T p.K507N Chr.1: 176043719-176043719 on Build GRCh38
6972731 c.1530G>T p.Q510H Chr.1: 176043710-176043710 on Build GRCh38
1295521 c.1517C>T p.S506L Chr.1: 176043723-176043723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17351808 Chr.1: 176043573-176043573 on Build GRCh38
rs1035281903 Chr.1: 176043597-176043597 on Build GRCh38
rs180880046 Chr.1: 176043673-176043673 on Build GRCh38
rs757642286 Chr.1: 176043699-176043699 on Build GRCh38
rs547649742 Chr.1: 176043658-176043658 on Build GRCh38
rs764312495 Chr.1: 176043681-176043681 on Build GRCh38
rs4233165 Chr.1: 176043671-176043671 on Build GRCh38
rs1056341983 Chr.1: 176043567-176043567 on Build GRCh38
rs1041964373 Chr.1: 176043601-176043601 on Build GRCh38
rs138078582 Chr.1: 176043648-176043648 on Build GRCh38
rs765550607 Chr.1: 176043672-176043672 on Build GRCh38
rs950379264 Chr.1: 176043574-176043574 on Build GRCh38
rs984170644 Chr.1: 176043540-176043540 on Build GRCh38
rs377195108 Chr.1: 176043675-176043675 on Build GRCh38
rs752134478 Chr.1: 176043689-176043689 on Build GRCh38
rs902889269 Chr.1: 176043641-176043641 on Build GRCh38
rs1028269251 Chr.1: 176043732-176043732 on Build GRCh38
rs754037094 Chr.1: 176043552-176043552 on Build GRCh38
rs150782230 Chr.1: 176043566-176043566 on Build GRCh38
rs113197099 Chr.1: 176043544-176043544 on Build GRCh38
rs951205331 Chr.1: 176043617-176043617 on Build GRCh38

View Full Product Details