Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101200645-101200916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOGAT3
Entrez Gene ID
346606
Gene Name
monoacylglycerol O-acyltransferase 3
Gene Aliases
DC7, DGAT2L2, MGAT3
UniGene
Hs.512217
Gene Chromosome Location
Chr.7: 101192886-101201131 on Build GRCh38
Transcripts
NM_001287147.1 NM_178176.3 XM_005250309.3 XM_011516126.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1292388 c.75C>T p.G25G Chr.7: 101200780-101200780 on Build GRCh38
4868140 c.109G>T p.G37C Chr.7: 101200746-101200746 on Build GRCh38
3366695 c.24G>A p.Q8Q Chr.7: 101200831-101200831 on Build GRCh38
5534634 c.55C>T p.Q19* Chr.7: 101200800-101200800 on Build GRCh38
5534635 c.55C>T p.Q19* Chr.7: 101200800-101200800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12674140 Chr.7: 101200730-101200730 on Build GRCh38
rs368413360 Chr.7: 101200728-101200728 on Build GRCh38
rs534888702 Chr.7: 101200795-101200795 on Build GRCh38
rs991534305 Chr.7: 101200668-101200668 on Build GRCh38
rs750103776 Chr.7: 101200742-101200742 on Build GRCh38
rs979438438 Chr.7: 101200688-101200689 on Build GRCh38
rs779115458 Chr.7: 101200896-101200896 on Build GRCh38
rs370642798 Chr.7: 101200704-101200704 on Build GRCh38
rs374350476 Chr.7: 101200711-101200711 on Build GRCh38
rs776324021 Chr.7: 101200736-101200736 on Build GRCh38
rs1055880719 Chr.7: 101200693-101200693 on Build GRCh38
rs772597129 Chr.7: 101200862-101200862 on Build GRCh38
rs767938339 Chr.7: 101200762-101200762 on Build GRCh38
rs768535153 Chr.7: 101200808-101200808 on Build GRCh38
rs577394565 Chr.7: 101200769-101200769 on Build GRCh38
rs1022651940 Chr.7: 101200690-101200690 on Build GRCh38
rs764606968 Chr.7: 101200741-101200741 on Build GRCh38
rs374109699 Chr.7: 101200733-101200733 on Build GRCh38
rs749717392 Chr.7: 101200780-101200780 on Build GRCh38
rs757922450 Chr.7: 101200749-101200749 on Build GRCh38
rs575094769 Chr.7: 101200824-101200824 on Build GRCh38
rs748659172 Chr.7: 101200699-101200699 on Build GRCh38
rs367702904 Chr.7: 101200892-101200892 on Build GRCh38
rs199850170 Chr.7: 101200885-101200885 on Build GRCh38
rs781478083 Chr.7: 101200696-101200696 on Build GRCh38
rs148580757 Chr.7: 101200782-101200782 on Build GRCh38
rs1009292081 Chr.7: 101200844-101200844 on Build GRCh38
rs762728129 Chr.7: 101200866-101200866 on Build GRCh38
rs944156776 Chr.7: 101200760-101200760 on Build GRCh38
rs756055308 Chr.7: 101200723-101200724 on Build GRCh38
rs772002514 Chr.7: 101200729-101200729 on Build GRCh38
rs371421116 Chr.7: 101200893-101200893 on Build GRCh38
rs778221176 Chr.7: 101200779-101200779 on Build GRCh38
rs765995401 Chr.7: 101200868-101200868 on Build GRCh38
rs761420827 Chr.7: 101200737-101200737 on Build GRCh38
rs756660816 Chr.7: 101200770-101200770 on Build GRCh38
rs772416358 Chr.7: 101200790-101200790 on Build GRCh38
rs548209628 Chr.7: 101200670-101200670 on Build GRCh38
rs553668394 Chr.7: 101200719-101200719 on Build GRCh38
rs753097625 Chr.7: 101200873-101200873 on Build GRCh38
rs771097906 Chr.7: 101200715-101200715 on Build GRCh38
rs746771595 Chr.7: 101200800-101200800 on Build GRCh38
rs746104352 Chr.7: 101200786-101200786 on Build GRCh38
rs937342862 Chr.7: 101200691-101200691 on Build GRCh38
rs141091754 Chr.7: 101200838-101200838 on Build GRCh38
rs376454972 Chr.7: 101200702-101200702 on Build GRCh38
rs1020381617 Chr.7: 101200883-101200883 on Build GRCh38
rs761598820 Chr.7: 101200837-101200837 on Build GRCh38

View Full Product Details