Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19346371-19346644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDHA1
Entrez Gene ID
5160
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 1
Gene Aliases
PDHA, PDHAD, PDHCE1A, PHE1A
UniGene
Hs.530331
Gene Chromosome Location
Chr.X: 19343893-19361707 on Build GRCh38
Transcripts
NM_000284.3 NM_001173454.1 NM_001173455.1 NM_001173456.1 XM_017029574.1

SNP Detail

SNP ID Chromosome Location
rs1003060579 Chr.X: 19346487-19346488 on Build GRCh38
rs145756935 Chr.X: 19346397-19346397 on Build GRCh38
rs891218284 Chr.X: 19346613-19346613 on Build GRCh38
rs751469147 Chr.X: 19346607-19346607 on Build GRCh38
rs753113002 Chr.X: 19346510-19346511 on Build GRCh38
rs145749914 Chr.X: 19346576-19346576 on Build GRCh38
rs765803649 Chr.X: 19346495-19346495 on Build GRCh38
rs1008772574 Chr.X: 19346617-19346617 on Build GRCh38
rs971633023 Chr.X: 19346493-19346493 on Build GRCh38
rs780276301 Chr.X: 19346558-19346558 on Build GRCh38
rs1054327928 Chr.X: 19346527-19346527 on Build GRCh38
rs750462643 Chr.X: 19346514-19346514 on Build GRCh38
rs760713655 Chr.X: 19346494-19346494 on Build GRCh38
rs1006433666 Chr.X: 19346601-19346601 on Build GRCh38
rs1050263814 Chr.X: 19346604-19346604 on Build GRCh38
rs754393427 Chr.X: 19346494-19346495 on Build GRCh38
rs7883320 Chr.X: 19346526-19346526 on Build GRCh38
rs756180699 Chr.X: 19346455-19346455 on Build GRCh38
rs766213373 Chr.X: 19346588-19346588 on Build GRCh38
rs915705058 Chr.X: 19346467-19346467 on Build GRCh38
rs182962186 Chr.X: 19346575-19346575 on Build GRCh38
rs747051654 Chr.X: 19346599-19346599 on Build GRCh38
rs1027338367 Chr.X: 19346394-19346394 on Build GRCh38

View Full Product Details