Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74165201-74165426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB1A
Entrez Gene ID
55233
Gene Name
MOB kinase activator 1A
Gene Aliases
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B
UniGene
Hs.602092
Gene Chromosome Location
Chr.2: 74152598-74178967 on Build GRCh38
Transcripts
NM_001317110.1 NM_001317111.1 NM_001317112.1 NM_018221.4 XM_011532948.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3372870 c.317C>T p.P106L Chr.2: 74165310-74165310 on Build GRCh38
6398671 c.284A>T p.Y95F Chr.2: 74165343-74165343 on Build GRCh38
6398672 c.284A>T p.Y95F Chr.2: 74165343-74165343 on Build GRCh38
6578002 c.391C>G p.L131V Chr.2: 74165236-74165236 on Build GRCh38
6578003 c.391C>G p.L131V Chr.2: 74165236-74165236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777158152 Chr.2: 74165267-74165267 on Build GRCh38
rs771497183 Chr.2: 74165264-74165264 on Build GRCh38
rs923721875 Chr.2: 74165369-74165369 on Build GRCh38
rs995323977 Chr.2: 74165231-74165231 on Build GRCh38
rs764167328 Chr.2: 74165358-74165358 on Build GRCh38
rs377622653 Chr.2: 74165393-74165393 on Build GRCh38
rs753060505 Chr.2: 74165338-74165338 on Build GRCh38
rs763243857 Chr.2: 74165348-74165348 on Build GRCh38
rs954568506 Chr.2: 74165308-74165308 on Build GRCh38
rs370037758 Chr.2: 74165374-74165374 on Build GRCh38
rs765651367 Chr.2: 74165282-74165282 on Build GRCh38
rs760009975 Chr.2: 74165268-74165268 on Build GRCh38
rs751683278 Chr.2: 74165367-74165367 on Build GRCh38
rs1028796140 Chr.2: 74165285-74165285 on Build GRCh38
rs760054125 Chr.2: 74165237-74165237 on Build GRCh38

View Full Product Details