Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125547519-125547774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSBPL11
Entrez Gene ID
114885
Gene Name
oxysterol binding protein like 11
Gene Aliases
ORP-11, ORP11, OSBP12, TCCCIA00292
UniGene
Hs.477440
Gene Chromosome Location
Chr.3: 125528858-125595537 on Build GRCh38
Transcripts
NM_022776.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1037669 c.1692A>G p.T564T Chr.3: 125547555-125547555 on Build GRCh38
5650054 c.1683_1685delAGA p.E562delE Chr.3: 125547562-125547564 on Build GRCh38
6735649 c.1674G>A p.E558E Chr.3: 125547573-125547573 on Build GRCh38
6735650 c.1675C>T p.H559Y Chr.3: 125547572-125547572 on Build GRCh38
1693394 c.1702C>T p.P568S Chr.3: 125547545-125547545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35535572 Chr.3: 125547543-125547543 on Build GRCh38
rs376930244 Chr.3: 125547590-125547590 on Build GRCh38
rs745562644 Chr.3: 125547701-125547701 on Build GRCh38
rs545111926 Chr.3: 125547698-125547698 on Build GRCh38
rs916847464 Chr.3: 125547681-125547681 on Build GRCh38
rs977843996 Chr.3: 125547635-125547635 on Build GRCh38
rs918757804 Chr.3: 125547704-125547704 on Build GRCh38
rs771344356 Chr.3: 125547608-125547608 on Build GRCh38
rs377669724 Chr.3: 125547644-125547644 on Build GRCh38
rs1037895768 Chr.3: 125547736-125547736 on Build GRCh38
rs776960822 Chr.3: 125547611-125547611 on Build GRCh38
rs1032086812 Chr.3: 125547661-125547661 on Build GRCh38
rs949676626 Chr.3: 125547696-125547696 on Build GRCh38
rs940880703 Chr.3: 125547699-125547699 on Build GRCh38
rs757897046 Chr.3: 125547705-125547705 on Build GRCh38
rs191358415 Chr.3: 125547633-125547633 on Build GRCh38
rs757098659 Chr.3: 125547563-125547563 on Build GRCh38
rs759452401 Chr.3: 125547620-125547620 on Build GRCh38
rs990481452 Chr.3: 125547667-125547667 on Build GRCh38
rs954376471 Chr.3: 125547656-125547656 on Build GRCh38
rs532130436 Chr.3: 125547542-125547542 on Build GRCh38
rs773285359 Chr.3: 125547570-125547570 on Build GRCh38
rs957729217 Chr.3: 125547662-125547662 on Build GRCh38

View Full Product Details