Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141476597-141476852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141430591-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGC3
Entrez Gene ID
5098
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
PC43, PCDH-GAMMA-C3, PCDH2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141476002-141512977 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4609799 c.504_505delTT p.L169fs*4 Chr.5: 141476620-141476621 on Build GRCh38
377265 c.547C>T p.R183C Chr.5: 141476663-141476663 on Build GRCh38
6102577 c.536A>G p.Y179C Chr.5: 141476652-141476652 on Build GRCh38
4712639 c.601G>A p.A201T Chr.5: 141476717-141476717 on Build GRCh38
1671980 c.616C>T p.R206W Chr.5: 141476732-141476732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138480390 Chr.5: 141476686-141476686 on Build GRCh38
rs1015622831 Chr.5: 141476742-141476742 on Build GRCh38
rs146188020 Chr.5: 141476714-141476714 on Build GRCh38
rs766227340 Chr.5: 141476791-141476791 on Build GRCh38
rs777922517 Chr.5: 141476808-141476808 on Build GRCh38
rs749347013 Chr.5: 141476739-141476739 on Build GRCh38
rs773763605 Chr.5: 141476631-141476631 on Build GRCh38
rs761650380 Chr.5: 141476683-141476684 on Build GRCh38
rs201463036 Chr.5: 141476780-141476780 on Build GRCh38
rs185280755 Chr.5: 141476824-141476824 on Build GRCh38
rs752954707 Chr.5: 141476801-141476801 on Build GRCh38
rs769909773 Chr.5: 141476732-141476732 on Build GRCh38
rs771355583 Chr.5: 141476752-141476752 on Build GRCh38
rs2075661 Chr.5: 141476698-141476698 on Build GRCh38
rs754785656 Chr.5: 141476793-141476793 on Build GRCh38
rs982396841 Chr.5: 141476777-141476777 on Build GRCh38
rs762236731 Chr.5: 141476727-141476727 on Build GRCh38
rs754731820 Chr.5: 141476704-141476704 on Build GRCh38
rs759887729 Chr.5: 141476657-141476657 on Build GRCh38
rs756358461 Chr.5: 141476803-141476803 on Build GRCh38
rs199871912 Chr.5: 141476715-141476715 on Build GRCh38
rs115808055 Chr.5: 141476782-141476782 on Build GRCh38
rs746365316 Chr.5: 141476833-141476833 on Build GRCh38
rs997136356 Chr.5: 141476638-141476638 on Build GRCh38
rs190269177 Chr.5: 141476656-141476656 on Build GRCh38
rs751321222 Chr.5: 141476702-141476702 on Build GRCh38
rs779348525 Chr.5: 141476831-141476831 on Build GRCh38
rs994726301 Chr.5: 141476632-141476632 on Build GRCh38
rs373439335 Chr.5: 141476733-141476733 on Build GRCh38
rs899753438 Chr.5: 141476628-141476628 on Build GRCh38
rs150444699 Chr.5: 141476645-141476645 on Build GRCh38
rs570982273 Chr.5: 141476764-141476764 on Build GRCh38
rs376910320 Chr.5: 141476792-141476792 on Build GRCh38
rs762462741 Chr.5: 141476789-141476789 on Build GRCh38
rs753538822 Chr.5: 141476676-141476676 on Build GRCh38
rs764674164 Chr.5: 141476776-141476776 on Build GRCh38
rs758302668 Chr.5: 141476697-141476697 on Build GRCh38
rs771366262 Chr.5: 141476646-141476646 on Build GRCh38
rs201538255 Chr.5: 141476796-141476796 on Build GRCh38
rs774243572 Chr.5: 141476680-141476681 on Build GRCh38
rs112156044 Chr.5: 141476771-141476771 on Build GRCh38
rs750429892 Chr.5: 141476696-141476696 on Build GRCh38
rs182518072 Chr.5: 141476658-141476658 on Build GRCh38
rs749333814 Chr.5: 141476813-141476813 on Build GRCh38
rs368919360 Chr.5: 141476820-141476820 on Build GRCh38
rs987205396 Chr.5: 141476682-141476682 on Build GRCh38
rs369748404 Chr.5: 141476671-141476671 on Build GRCh38

View Full Product Details