Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112278226-112278451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIFA
Entrez Gene ID
92610
Gene Name
TRAF interacting protein with forkhead associated domain
Gene Aliases
T2BP, T6BP, TIFAA
UniGene
Hs.310640
Gene Chromosome Location
Chr.4: 112275626-112285903 on Build GRCh38
Transcripts
NM_052864.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471599 c.100A>G p.I34V Chr.4: 112278317-112278317 on Build GRCh38
3824965 c.89T>G p.I30R Chr.4: 112278328-112278328 on Build GRCh38
51003 c.129C>T p.S43S Chr.4: 112278288-112278288 on Build GRCh38
1579227 c.133G>A p.E45K Chr.4: 112278284-112278284 on Build GRCh38
3824966 c.86G>C p.G29A Chr.4: 112278331-112278331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147617925 Chr.4: 112278263-112278263 on Build GRCh38
rs767562614 Chr.4: 112278406-112278406 on Build GRCh38
rs745727348 Chr.4: 112278276-112278276 on Build GRCh38
rs778091662 Chr.4: 112278278-112278278 on Build GRCh38
rs762733411 Chr.4: 112278370-112278370 on Build GRCh38
rs6834237 Chr.4: 112278361-112278361 on Build GRCh38
rs369263822 Chr.4: 112278420-112278420 on Build GRCh38
rs942451428 Chr.4: 112278285-112278285 on Build GRCh38
rs774739590 Chr.4: 112278314-112278314 on Build GRCh38
rs766122633 Chr.4: 112278376-112278376 on Build GRCh38
rs771122276 Chr.4: 112278288-112278288 on Build GRCh38
rs373367609 Chr.4: 112278425-112278425 on Build GRCh38
rs759343955 Chr.4: 112278384-112278384 on Build GRCh38
rs984353886 Chr.4: 112278317-112278317 on Build GRCh38
rs765477260 Chr.4: 112278309-112278310 on Build GRCh38
rs568944599 Chr.4: 112278268-112278268 on Build GRCh38
rs973873639 Chr.4: 112278306-112278306 on Build GRCh38
rs758318779 Chr.4: 112278405-112278405 on Build GRCh38
rs750371054 Chr.4: 112278349-112278349 on Build GRCh38
rs1030595375 Chr.4: 112278260-112278260 on Build GRCh38
rs545028676 Chr.4: 112278397-112278397 on Build GRCh38
rs761391882 Chr.4: 112278360-112278360 on Build GRCh38
rs749425151 Chr.4: 112278283-112278283 on Build GRCh38
rs938463738 Chr.4: 112278411-112278411 on Build GRCh38
rs537549372 Chr.4: 112278378-112278378 on Build GRCh38
rs907123851 Chr.4: 112278409-112278409 on Build GRCh38
rs772755702 Chr.4: 112278366-112278366 on Build GRCh38
rs200608467 Chr.4: 112278266-112278266 on Build GRCh38
rs775803812 Chr.4: 112278347-112278347 on Build GRCh38
rs748270021 Chr.4: 112278265-112278265 on Build GRCh38
rs746228776 Chr.4: 112278335-112278335 on Build GRCh38
rs772627267 Chr.4: 112278342-112278342 on Build GRCh38

View Full Product Details