Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41308680-41308937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFRP1
Entrez Gene ID
6422
Gene Name
secreted frizzled related protein 1
Gene Aliases
FRP, FRP-1, FRP1, FrzA, SARP2
UniGene
Hs.213424
Gene Chromosome Location
Chr.8: 41261957-41309471 on Build GRCh38
Transcripts
NM_003012.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258097 c.454C>A p.P152T Chr.8: 41308706-41308706 on Build GRCh38
4650807 c.281C>T p.A94V Chr.8: 41308879-41308879 on Build GRCh38
88663 c.355C>G p.P119A Chr.8: 41308805-41308805 on Build GRCh38
6312110 c.383C>A p.P128Q Chr.8: 41308777-41308777 on Build GRCh38
1673868 c.410G>A p.R137H Chr.8: 41308750-41308750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374492383 Chr.8: 41308903-41308903 on Build GRCh38
rs373338351 Chr.8: 41308755-41308755 on Build GRCh38
rs965639106 Chr.8: 41308731-41308731 on Build GRCh38
rs572275992 Chr.8: 41308780-41308780 on Build GRCh38
rs370348984 Chr.8: 41308841-41308841 on Build GRCh38
rs766649756 Chr.8: 41308776-41308776 on Build GRCh38
rs768211131 Chr.8: 41308735-41308735 on Build GRCh38
rs76486417 Chr.8: 41308916-41308916 on Build GRCh38
rs779643083 Chr.8: 41308856-41308856 on Build GRCh38
rs535023429 Chr.8: 41308790-41308790 on Build GRCh38
rs746645058 Chr.8: 41308863-41308863 on Build GRCh38
rs750147715 Chr.8: 41308840-41308840 on Build GRCh38
rs746562361 Chr.8: 41308733-41308733 on Build GRCh38
rs774962972 Chr.8: 41308839-41308839 on Build GRCh38
rs75563193 Chr.8: 41308849-41308849 on Build GRCh38
rs137876408 Chr.8: 41308823-41308823 on Build GRCh38
rs1057451989 Chr.8: 41308891-41308891 on Build GRCh38
rs937435721 Chr.8: 41308880-41308880 on Build GRCh38
rs558728015 Chr.8: 41308743-41308743 on Build GRCh38
rs753766190 Chr.8: 41308809-41308809 on Build GRCh38
rs759598692 Chr.8: 41308785-41308785 on Build GRCh38
rs745540217 Chr.8: 41308711-41308711 on Build GRCh38
rs773726937 Chr.8: 41308907-41308907 on Build GRCh38
rs918768957 Chr.8: 41308777-41308777 on Build GRCh38
rs768103490 Chr.8: 41308876-41308876 on Build GRCh38
rs942441359 Chr.8: 41308734-41308734 on Build GRCh38
rs762360499 Chr.8: 41308744-41308744 on Build GRCh38
rs757134616 Chr.8: 41308833-41308833 on Build GRCh38
rs780705476 Chr.8: 41308878-41308878 on Build GRCh38
rs771668038 Chr.8: 41308715-41308715 on Build GRCh38
rs748698728 Chr.8: 41308892-41308892 on Build GRCh38
rs779616580 Chr.8: 41308726-41308726 on Build GRCh38

View Full Product Details