Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54930496-54930731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCR1
Entrez Gene ID
9437
Gene Name
natural cytotoxicity triggering receptor 1
Gene Aliases
CD335, LY94, NK-p46, NKP46
UniGene
Hs.97084
Gene Chromosome Location
Chr.19: 54906042-54938211 on Build GRCh38
Transcripts
XM_011527530.2 NM_001127255.1 NM_139176.3 NM_206828.3 XM_006723075.3 XM_006723076.3 XM_011526596.2 XM_011526599.2 XM_011526601.2

Gene Symbol
NLRP7
Entrez Gene ID
199713
Gene Name
NLR family pyrin domain containing 7
Gene Aliases
CLR19.4, HYDM, NALP7, NOD12, PAN7, PYPAF3
UniGene
Hs.351118
Gene Chromosome Location
Chr.19: 54923509-54965184 on Build GRCh38
Transcripts
XM_011527530.2 NM_001127255.1 NM_139176.3 NM_206828.3 XM_006723075.3 XM_006723076.3 XM_011526596.2 XM_011526599.2 XM_011526601.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209054 c.2658G>A p.Q886Q Chr.19: 54930567-54930567 on Build GRCh38
3681061 c.2691A>T p.A897A Chr.19: 54930534-54930534 on Build GRCh38
6489816 c.2675G>A p.C892Y Chr.19: 54930634-54930634 on Build GRCh38
3681062 c.2859A>T p.A953A Chr.19: 54930534-54930534 on Build GRCh38
7160196 c.2680C>T p.L894F Chr.19: 54930545-54930545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868526422 Chr.19: 54930523-54930523 on Build GRCh38
rs104895503 Chr.19: 54930571-54930571 on Build GRCh38
rs781125532 Chr.19: 54930535-54930535 on Build GRCh38
rs768641374 Chr.19: 54930592-54930592 on Build GRCh38
rs61746780 Chr.19: 54930603-54930603 on Build GRCh38
rs374729905 Chr.19: 54930615-54930615 on Build GRCh38
rs371132510 Chr.19: 54930605-54930605 on Build GRCh38
rs779014649 Chr.19: 54930596-54930596 on Build GRCh38
rs748286893 Chr.19: 54930704-54930704 on Build GRCh38
rs780748394 Chr.19: 54930619-54930619 on Build GRCh38
rs1025505024 Chr.19: 54930657-54930657 on Build GRCh38
rs777649835 Chr.19: 54930689-54930689 on Build GRCh38
rs778959473 Chr.19: 54930693-54930693 on Build GRCh38
rs753986364 Chr.19: 54930671-54930671 on Build GRCh38
rs775039016 Chr.19: 54930616-54930616 on Build GRCh38
rs755890731 Chr.19: 54930662-54930662 on Build GRCh38
rs780272212 Chr.19: 54930668-54930668 on Build GRCh38
rs371741023 Chr.19: 54930559-54930559 on Build GRCh38
rs1018466180 Chr.19: 54930579-54930579 on Build GRCh38
rs1030968324 Chr.19: 54930618-54930618 on Build GRCh38
rs761824633 Chr.19: 54930641-54930641 on Build GRCh38
rs762712137 Chr.19: 54930624-54930624 on Build GRCh38
rs750297430 Chr.19: 54930543-54930543 on Build GRCh38
rs755270258 Chr.19: 54930672-54930672 on Build GRCh38
rs269950 Chr.19: 54930534-54930534 on Build GRCh38
rs932369211 Chr.19: 54930549-54930549 on Build GRCh38
rs372626501 Chr.19: 54930654-54930654 on Build GRCh38
rs269951 Chr.19: 54930627-54930627 on Build GRCh38
rs748623991 Chr.19: 54930601-54930601 on Build GRCh38
rs368560112 Chr.19: 54930630-54930630 on Build GRCh38
rs756014953 Chr.19: 54930642-54930642 on Build GRCh38
rs867728573 Chr.19: 54930652-54930652 on Build GRCh38
rs61750468 Chr.19: 54930588-54930588 on Build GRCh38

View Full Product Details