Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46065058-46065283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA5
Entrez Gene ID
57727
Gene Name
nuclear receptor coactivator 5
Gene Aliases
CIA, bA465L10.6
UniGene
Hs.654991
Gene Chromosome Location
Chr.20: 46060985-46089962 on Build GRCh38
Transcripts
NM_020967.2 XM_005260474.3 XM_011528951.2 XM_017027988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6331875 c.669G>A p.L223L Chr.20: 46065189-46065189 on Build GRCh38
6334109 c.792C>A p.R264R Chr.20: 46065066-46065066 on Build GRCh38
4403915 c.752C>T p.P251L Chr.20: 46065106-46065106 on Build GRCh38
478231 c.650G>T p.G217V Chr.20: 46065208-46065208 on Build GRCh38
7179599 c.727G>C p.E243Q Chr.20: 46065131-46065131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369442369 Chr.20: 46065156-46065156 on Build GRCh38
rs768417079 Chr.20: 46065205-46065205 on Build GRCh38
rs117464940 Chr.20: 46065189-46065189 on Build GRCh38
rs375379866 Chr.20: 46065129-46065129 on Build GRCh38
rs533531058 Chr.20: 46065153-46065153 on Build GRCh38
rs754214998 Chr.20: 46065078-46065078 on Build GRCh38
rs576890583 Chr.20: 46065213-46065213 on Build GRCh38
rs778860316 Chr.20: 46065180-46065180 on Build GRCh38
rs17450969 Chr.20: 46065258-46065258 on Build GRCh38
rs746293216 Chr.20: 46065235-46065238 on Build GRCh38
rs911121917 Chr.20: 46065105-46065105 on Build GRCh38
rs147641777 Chr.20: 46065098-46065098 on Build GRCh38
rs763431816 Chr.20: 46065243-46065243 on Build GRCh38
rs760317447 Chr.20: 46065196-46065196 on Build GRCh38
rs749046730 Chr.20: 46065171-46065171 on Build GRCh38
rs993297932 Chr.20: 46065112-46065112 on Build GRCh38
rs140004936 Chr.20: 46065247-46065247 on Build GRCh38
rs745822989 Chr.20: 46065182-46065182 on Build GRCh38
rs758436976 Chr.20: 46065109-46065109 on Build GRCh38
rs755056177 Chr.20: 46065249-46065249 on Build GRCh38
rs369032369 Chr.20: 46065093-46065093 on Build GRCh38
rs762176260 Chr.20: 46065219-46065219 on Build GRCh38
rs765556233 Chr.20: 46065239-46065239 on Build GRCh38
rs543923160 Chr.20: 46065244-46065244 on Build GRCh38
rs998871658 Chr.20: 46065092-46065092 on Build GRCh38
rs750843979 Chr.20: 46065241-46065241 on Build GRCh38
rs770646101 Chr.20: 46065174-46065174 on Build GRCh38
rs373151968 Chr.20: 46065191-46065191 on Build GRCh38
rs554043179 Chr.20: 46065087-46065087 on Build GRCh38
rs374572190 Chr.20: 46065261-46065261 on Build GRCh38
rs751690591 Chr.20: 46065135-46065135 on Build GRCh38
rs781418226 Chr.20: 46065257-46065257 on Build GRCh38

View Full Product Details