Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165727934-165728207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO1
Entrez Gene ID
54499
Gene Name
transmembrane and coiled-coil domains 1
Gene Aliases
HP10122, PCIA3, PNAS-136, TMCC4
UniGene
Hs.31498
Gene Chromosome Location
Chr.1: 165724291-165768922 on Build GRCh38
Transcripts
NM_001256164.1 NM_001256165.1 NM_019026.4 NR_045818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5029477 c.655C>T p.R219* Chr.1: 165728088-165728088 on Build GRCh38
1689076 c.548C>T p.P183L Chr.1: 165728042-165728042 on Build GRCh38
4852960 c.469-1G>A p.? Chr.1: 165728122-165728122 on Build GRCh38
1689075 c.701C>T p.P234L Chr.1: 165728042-165728042 on Build GRCh38
3671484 c.563C>T p.S188F Chr.1: 165728027-165728027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938095771 Chr.1: 165728062-165728062 on Build GRCh38
rs760783276 Chr.1: 165727961-165727961 on Build GRCh38
rs777739591 Chr.1: 165728044-165728045 on Build GRCh38
rs369419998 Chr.1: 165728166-165728166 on Build GRCh38
rs765006399 Chr.1: 165728000-165728000 on Build GRCh38
rs749040101 Chr.1: 165728168-165728170 on Build GRCh38
rs754672708 Chr.1: 165728153-165728153 on Build GRCh38
rs749769560 Chr.1: 165728049-165728049 on Build GRCh38
rs758217263 Chr.1: 165728019-165728019 on Build GRCh38
rs112969816 Chr.1: 165728040-165728040 on Build GRCh38
rs201918820 Chr.1: 165728169-165728169 on Build GRCh38
rs915211345 Chr.1: 165728103-165728103 on Build GRCh38
rs764294913 Chr.1: 165727970-165727970 on Build GRCh38
rs754021967 Chr.1: 165728135-165728135 on Build GRCh38
rs78363884 Chr.1: 165728104-165728104 on Build GRCh38
rs757302422 Chr.1: 165727984-165727984 on Build GRCh38
rs779929531 Chr.1: 165728147-165728147 on Build GRCh38
rs199592544 Chr.1: 165728145-165728145 on Build GRCh38
rs1042955590 Chr.1: 165728154-165728154 on Build GRCh38
rs528475298 Chr.1: 165728158-165728158 on Build GRCh38
rs752091077 Chr.1: 165728016-165728016 on Build GRCh38
rs1007306216 Chr.1: 165728184-165728184 on Build GRCh38
rs375742777 Chr.1: 165728137-165728137 on Build GRCh38
rs746684690 Chr.1: 165728032-165728032 on Build GRCh38
rs774768773 Chr.1: 165728071-165728071 on Build GRCh38
rs551954100 Chr.1: 165728165-165728165 on Build GRCh38
rs751181050 Chr.1: 165728152-165728152 on Build GRCh38
rs201904119 Chr.1: 165728088-165728088 on Build GRCh38
rs771396376 Chr.1: 165728064-165728064 on Build GRCh38
rs753789740 Chr.1: 165727976-165727976 on Build GRCh38
rs775529806 Chr.1: 165728083-165728083 on Build GRCh38
rs759757338 Chr.1: 165728078-165728078 on Build GRCh38
rs371420999 Chr.1: 165728136-165728136 on Build GRCh38
rs750387757 Chr.1: 165728138-165728138 on Build GRCh38
rs890247383 Chr.1: 165728181-165728181 on Build GRCh38
rs780718220 Chr.1: 165728038-165728038 on Build GRCh38
rs150796970 Chr.1: 165728082-165728082 on Build GRCh38
rs768112478 Chr.1: 165728033-165728033 on Build GRCh38
rs928077940 Chr.1: 165728021-165728021 on Build GRCh38
rs1051578144 Chr.1: 165728177-165728177 on Build GRCh38
rs779514586 Chr.1: 165728031-165728031 on Build GRCh38

View Full Product Details