Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 89328725-89328990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SECISBP2
Entrez Gene ID
79048
Gene Name
SECIS binding protein 2
Gene Aliases
SBP2
UniGene
Hs.59804
Gene Chromosome Location
Chr.9: 89318473-89367117 on Build GRCh38
Transcripts
NM_001282688.1 NM_001282689.1 NM_001282690.1 NM_024077.4 XM_005252196.2 XM_006717282.2 XM_011519000.2 XM_011519001.1 XM_011519002.1 XM_011519003.1 XM_017015122.1 XM_017015123.1 XM_017015124.1 XM_017015125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5876539 c.656C>T p.T219I Chr.9: 89328741-89328741 on Build GRCh38
7214922 c.764T>A p.L255Q Chr.9: 89328849-89328849 on Build GRCh38
81691 c.768A>G p.R256R Chr.9: 89328853-89328853 on Build GRCh38
1463505 c.646T>G p.F216V Chr.9: 89328731-89328731 on Build GRCh38
6116666 c.769G>C p.E257Q Chr.9: 89328854-89328854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756038364 Chr.9: 89328850-89328850 on Build GRCh38
rs201068154 Chr.9: 89328849-89328849 on Build GRCh38
rs765529994 Chr.9: 89328844-89328844 on Build GRCh38
rs752638506 Chr.9: 89328783-89328783 on Build GRCh38
rs142217806 Chr.9: 89328754-89328754 on Build GRCh38
rs192655660 Chr.9: 89328866-89328866 on Build GRCh38
rs768996103 Chr.9: 89328836-89328836 on Build GRCh38
rs374199098 Chr.9: 89328897-89328897 on Build GRCh38
rs949561573 Chr.9: 89328892-89328892 on Build GRCh38
rs771636918 Chr.9: 89328833-89328833 on Build GRCh38
rs994897551 Chr.9: 89328855-89328855 on Build GRCh38
rs950378382 Chr.9: 89328755-89328755 on Build GRCh38
rs781188422 Chr.9: 89328782-89328782 on Build GRCh38
rs531603787 Chr.9: 89328809-89328809 on Build GRCh38
rs750744951 Chr.9: 89328847-89328847 on Build GRCh38
rs76202187 Chr.9: 89328906-89328906 on Build GRCh38
rs1018845906 Chr.9: 89328970-89328970 on Build GRCh38
rs757083455 Chr.9: 89328873-89328873 on Build GRCh38
rs765892198 Chr.9: 89328813-89328813 on Build GRCh38
rs767286049 Chr.9: 89328816-89328824 on Build GRCh38
rs777575684 Chr.9: 89328858-89328858 on Build GRCh38
rs763275926 Chr.9: 89328932-89328932 on Build GRCh38
rs753536377 Chr.9: 89328835-89328835 on Build GRCh38
rs942261112 Chr.9: 89328845-89328845 on Build GRCh38
rs117852708 Chr.9: 89328969-89328969 on Build GRCh38
rs762268068 Chr.9: 89328904-89328904 on Build GRCh38
rs778585441 Chr.9: 89328816-89328816 on Build GRCh38
rs745577063 Chr.9: 89328881-89328881 on Build GRCh38
rs371846875 Chr.9: 89328756-89328756 on Build GRCh38
rs773002743 Chr.9: 89328828-89328829 on Build GRCh38
rs554140618 Chr.9: 89328882-89328882 on Build GRCh38
rs372551613 Chr.9: 89328898-89328898 on Build GRCh38
rs745350838 Chr.9: 89328818-89328818 on Build GRCh38
rs761804270 Chr.9: 89328799-89328799 on Build GRCh38
rs534481161 Chr.9: 89328880-89328880 on Build GRCh38
rs374879321 Chr.9: 89328781-89328781 on Build GRCh38
rs762128719 Chr.9: 89328766-89328766 on Build GRCh38
rs777436493 Chr.9: 89328805-89328805 on Build GRCh38
rs765364217 Chr.9: 89328770-89328770 on Build GRCh38
rs752912689 Chr.9: 89328907-89328908 on Build GRCh38
rs770344638 Chr.9: 89328912-89328912 on Build GRCh38
rs143420456 Chr.9: 89328784-89328784 on Build GRCh38
rs779432748 Chr.9: 89328891-89328891 on Build GRCh38
rs991499769 Chr.9: 89328874-89328874 on Build GRCh38
rs902640769 Chr.9: 89328904-89328905 on Build GRCh38
rs139203513 Chr.9: 89328838-89328838 on Build GRCh38
rs773512714 Chr.9: 89328913-89328913 on Build GRCh38
rs759715421 Chr.9: 89328862-89328862 on Build GRCh38
rs750548843 Chr.9: 89328780-89328780 on Build GRCh38
rs765520982 Chr.9: 89328901-89328904 on Build GRCh38

View Full Product Details