Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15041496-15041751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTAN1
Entrez Gene ID
123803
Gene Name
N-terminal asparagine amidase
Gene Aliases
PNAA, PNAD
UniGene
Hs.592045
Gene Chromosome Location
Chr.16: 15037853-15056079 on Build GRCh38
Transcripts
NM_001270766.1 NM_001270767.1 NM_173474.3 XM_011522355.2 NM_001285449.1 NM_001324020.1 NM_001324021.1 XM_017023065.1

Gene Symbol
PDXDC1
Entrez Gene ID
23042
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Gene Aliases
LP8165
UniGene
Hs.370781
Gene Chromosome Location
Chr.16: 14974591-15139410 on Build GRCh38
Transcripts
NM_001270766.1 NM_001270767.1 NM_173474.3 XM_011522355.2 NM_001285449.1 NM_001324020.1 NM_001324021.1 XM_017023065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3741780 c.487+3A>G p.? Chr.16: 15041620-15041620 on Build GRCh38
7197721 c.487G>A p.E163K Chr.16: 15041623-15041623 on Build GRCh38
5452450 c.476T>G p.L159* Chr.16: 15041634-15041634 on Build GRCh38
434678 c.442G>C p.D148H Chr.16: 15041668-15041668 on Build GRCh38
4340465 c.465C>G p.H155Q Chr.16: 15041645-15041645 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753091147 Chr.16: 15041725-15041725 on Build GRCh38
rs368411141 Chr.16: 15041642-15041642 on Build GRCh38
rs763182729 Chr.16: 15041731-15041731 on Build GRCh38
rs1040016171 Chr.16: 15041542-15041542 on Build GRCh38
rs367642054 Chr.16: 15041699-15041699 on Build GRCh38
rs376151338 Chr.16: 15041619-15041619 on Build GRCh38
rs377218066 Chr.16: 15041708-15041708 on Build GRCh38
rs372285060 Chr.16: 15041685-15041685 on Build GRCh38
rs925977498 Chr.16: 15041713-15041713 on Build GRCh38
rs761584490 Chr.16: 15041657-15041657 on Build GRCh38
rs769478424 Chr.16: 15041680-15041680 on Build GRCh38
rs79204324 Chr.16: 15041689-15041689 on Build GRCh38
rs927048805 Chr.16: 15041541-15041541 on Build GRCh38
rs781559830 Chr.16: 15041589-15041589 on Build GRCh38
rs141773371 Chr.16: 15041575-15041575 on Build GRCh38
rs753051788 Chr.16: 15041591-15041591 on Build GRCh38
rs777914622 Chr.16: 15041610-15041610 on Build GRCh38
rs945988067 Chr.16: 15041659-15041659 on Build GRCh38
rs374878101 Chr.16: 15041703-15041703 on Build GRCh38
rs200102960 Chr.16: 15041668-15041668 on Build GRCh38
rs565245548 Chr.16: 15041696-15041696 on Build GRCh38
rs201675903 Chr.16: 15041700-15041700 on Build GRCh38
rs111350492 Chr.16: 15041726-15041726 on Build GRCh38
rs572586323 Chr.16: 15041540-15041540 on Build GRCh38
rs768760001 Chr.16: 15041633-15041633 on Build GRCh38
rs561411971 Chr.16: 15041598-15041598 on Build GRCh38
rs780097202 Chr.16: 15041622-15041622 on Build GRCh38
rs958500069 Chr.16: 15041567-15041567 on Build GRCh38
rs554179142 Chr.16: 15041519-15041519 on Build GRCh38
rs746961668 Chr.16: 15041631-15041631 on Build GRCh38
rs371794878 Chr.16: 15041727-15041727 on Build GRCh38
rs755297376 Chr.16: 15041576-15041576 on Build GRCh38
rs755598115 Chr.16: 15041729-15041732 on Build GRCh38
rs1042152020 Chr.16: 15041596-15041596 on Build GRCh38
rs762385332 Chr.16: 15041697-15041697 on Build GRCh38
rs984712259 Chr.16: 15041520-15041520 on Build GRCh38
rs749329613 Chr.16: 15041611-15041611 on Build GRCh38

View Full Product Details