Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6693928-6694201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100P
Entrez Gene ID
6286
Gene Name
S100 calcium binding protein P
Gene Aliases
MIG9
UniGene
Hs.2962
Gene Chromosome Location
Chr.4: 6693839-6697170 on Build GRCh38
Transcripts
NM_005980.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1056666 c.20C>T p.A7V Chr.4: 6693952-6693952 on Build GRCh38
3604962 c.116A>C p.K39T Chr.4: 6694048-6694048 on Build GRCh38
5988877 c.114G>T p.E38D Chr.4: 6694046-6694046 on Build GRCh38
241505 c.56C>T p.S19L Chr.4: 6693988-6693988 on Build GRCh38
5717176 c.50G>A p.R17Q Chr.4: 6693982-6693982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921808639 Chr.4: 6694138-6694138 on Build GRCh38
rs750128716 Chr.4: 6694080-6694080 on Build GRCh38
rs4712 Chr.4: 6694046-6694046 on Build GRCh38
rs201438904 Chr.4: 6694101-6694101 on Build GRCh38
rs779322318 Chr.4: 6694020-6694020 on Build GRCh38
rs879176746 Chr.4: 6694071-6694071 on Build GRCh38
rs574641321 Chr.4: 6694009-6694009 on Build GRCh38
rs545595778 Chr.4: 6694126-6694126 on Build GRCh38
rs543945473 Chr.4: 6693981-6693981 on Build GRCh38
rs187031070 Chr.4: 6693972-6693972 on Build GRCh38
rs771396235 Chr.4: 6694040-6694040 on Build GRCh38
rs909031410 Chr.4: 6694160-6694160 on Build GRCh38
rs879028242 Chr.4: 6694074-6694074 on Build GRCh38
rs766298645 Chr.4: 6694082-6694082 on Build GRCh38
rs531054091 Chr.4: 6694164-6694164 on Build GRCh38
rs768130751 Chr.4: 6694050-6694050 on Build GRCh38
rs148427693 Chr.4: 6694030-6694030 on Build GRCh38
rs779089634 Chr.4: 6694085-6694085 on Build GRCh38
rs565304564 Chr.4: 6694129-6694129 on Build GRCh38
rs767306291 Chr.4: 6694059-6694059 on Build GRCh38
rs991472319 Chr.4: 6694043-6694043 on Build GRCh38
rs747927620 Chr.4: 6694152-6694152 on Build GRCh38
rs758632639 Chr.4: 6694099-6694099 on Build GRCh38
rs761417487 Chr.4: 6694058-6694058 on Build GRCh38
rs142529937 Chr.4: 6694034-6694034 on Build GRCh38
rs761044965 Chr.4: 6693967-6693967 on Build GRCh38
rs995888097 Chr.4: 6694141-6694141 on Build GRCh38
rs762410281 Chr.4: 6694044-6694044 on Build GRCh38
rs758463696 Chr.4: 6693966-6693966 on Build GRCh38
rs755084534 Chr.4: 6694019-6694019 on Build GRCh38
rs761470541 Chr.4: 6694118-6694118 on Build GRCh38
rs773785885 Chr.4: 6694113-6694113 on Build GRCh38
rs371875811 Chr.4: 6694083-6694083 on Build GRCh38
rs199731575 Chr.4: 6693988-6693988 on Build GRCh38
rs931886070 Chr.4: 6694151-6694151 on Build GRCh38
rs777786695 Chr.4: 6694165-6694165 on Build GRCh38
rs367657958 Chr.4: 6694081-6694081 on Build GRCh38
rs749386092 Chr.4: 6694017-6694017 on Build GRCh38
rs751181267 Chr.4: 6693989-6693989 on Build GRCh38
rs866006725 Chr.4: 6694024-6694024 on Build GRCh38
rs771770180 Chr.4: 6694119-6694119 on Build GRCh38
rs1021039536 Chr.4: 6693996-6693996 on Build GRCh38
rs908061986 Chr.4: 6694106-6694106 on Build GRCh38
rs141272313 Chr.4: 6694108-6694108 on Build GRCh38
rs746336288 Chr.4: 6694104-6694104 on Build GRCh38
rs755064985 Chr.4: 6694084-6694084 on Build GRCh38
rs747561018 Chr.4: 6694037-6694037 on Build GRCh38
rs139901528 Chr.4: 6693995-6693995 on Build GRCh38
rs757091503 Chr.4: 6693991-6693991 on Build GRCh38
rs146007022 Chr.4: 6693971-6693971 on Build GRCh38
rs1032934662 Chr.4: 6693982-6693982 on Build GRCh38
rs773641440 Chr.4: 6694036-6694036 on Build GRCh38
rs560138763 Chr.4: 6694092-6694092 on Build GRCh38
rs900142232 Chr.4: 6694135-6694135 on Build GRCh38
rs940527187 Chr.4: 6694161-6694161 on Build GRCh38
rs772536614 Chr.4: 6693949-6693949 on Build GRCh38
rs376830218 Chr.4: 6694100-6694100 on Build GRCh38
rs774012381 Chr.4: 6694055-6694055 on Build GRCh38
rs73089273 Chr.4: 6694007-6694007 on Build GRCh38
rs146198118 Chr.4: 6694012-6694012 on Build GRCh38
rs768183843 Chr.4: 6694105-6694105 on Build GRCh38
rs879100796 Chr.4: 6694072-6694072 on Build GRCh38

View Full Product Details